Product Image
Invitrogen™

pcDNA™6/TR vector Mammalian Expression Vector

The pcDNA™6⁄TR vector is part of the T-REx™ System (catalog K102001), which yields higher levels of induced expression than anyRead more
Have Questions?
Catalog NumberQuantity
V10252020 μg
Catalog number V102520
Price (USD)
846.00
Special offer
Online exclusive
Ends: 27-Jun-2026
900.00
Save 54.00 (6%)
20 µg
Quantity:
20 μg
Price (USD)
846.00
Special offer
Online exclusive
Ends: 27-Jun-2026
900.00
Save 54.00 (6%)
20 µg
The pcDNA™6⁄TR vector is part of the T-REx™ System (catalog K102001), which yields higher levels of induced expression than any other regulated mammalian expression system. It utilizes the complete CMV promoter and adds control elements from the bacterial tetracycline resistance operon to effectively repress and derepress transcription from one of the strongest mammalian promoter sequences known (1,2).

The regulatory vector, pcDNA™6⁄TR, is provided for high-level expression of the tetracycline repressor (TR) protein. This vector expresses the Blasticidin resistance gene for rapid selection of mammalian cell lines that stably express the TR protein.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemInducible
Delivery TypeTransfection
For Use With (Application)Regulated Expression
Product TypeMammalian Expression Vector
Quantity20 μg
Selection Agent (Eukaryotic)Blasticidin
Shipping ConditionRoom Temperature
VectorpcDNA
Product LineT-REx, pcDNA
PromoterCMV
Protein TagUntagged
Unit Size20 µg
Contents & Storage
20 μg of supercoiled vector (40 μL at 0.5 μg/μL) supplied in TE Buffer, pH 8.0. Store at -20°C.

Frequently asked questions (FAQs)

Can I use any Gateway entry vector to generate entry clones for use in RNAi applications?

No, you should use an entry vector that contains the elements necessary for RNA Polymerase III-dependent expression of your shRNA (i.e., Pol III promoter and terminator).

What is a dose response curve or kill curve? And can you outline the steps involved?

A dose response curve or kill curve is a simple method for determining the optimal antibiotic concentration to use when establishing a stable cell line. Untransfected cells are grown in a medium containing antibiotic at varying concentrations in order to determine the lowest amount of antibiotic needed to achieve complete cell death. The basic steps for performing a dose response curve or kill curve are as follows:

- Plate untransfected cells at 25% confluence, and grow them in a medium containing increasing concentrations of the antibiotic. For some antibiotics, you will need to calculate the amount of active drug to control for lot variation.
- Replenish the selective medium every 3-4 days. After 10-12 days, examine the dishes for viable cells. The cells may divide once or twice in the selective medium before cell death begins to occur.
- Look for the minimum concentration of antibiotic that resulted in complete cell death. This is the optimal antibiotic concentration to use for stable selection.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Can I create stable cell lines using pENTR/U6 entry vector or the pENTR/H1/TO vector?

Unfortunately, the pENTR/U6 vector does not contain a selection marker; therefore, only transient RNAi analysis may be performed. If you wish to generate stable cell lines, perform an LR reaction into an appropriate Gateway destination vector to generate expression clones.
The pENTR/H1/TO vector contains the Zeocin resistance gene to facilitate generation of cell lines that inducbily express the shRNA of interest. Perform a kill curve to determine the minimum concentration of Zeocin that is required to kill your untransfected mammalian cell line. Please note that Zeocin-sensitive cells do not round up and detach from the plate, but rather may increase in size, show abnormal cell shape, display presence of large empty vesicles in the cytoplasm, or show breakdown of plasma/nuclear membranes.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

What loop sequence should I use when designing my shRNA for cloning? Do you have any guidelines I should follow?

You can use a loop sequence of any length ranging from 4 to 11 nucleotides, although short loops (i.e., 4-7 nucleotides) are generally preferred. Avoid using a loop sequence containing thymidines (Ts), as they may cause early termination. This is particularly true if the target sequence itself ends in one or more T nucleotides. Here are some loop sequences we recommend:

- 5' - CGAA - 3'
- 5' - AACG - 3'
- 5' - GAGA - 3'

What considerations regarding transcription initiation should I take when designing my shRNA for cloning?

Transcription of the shRNA initiates at the first base following the end of the U6 promoter sequence. In the top-strand oligo, the transcription initiation site corresponds to the first nucleotide following the 4 bp CACC sequence added to permit directional cloning. We recommend initiating the shRNA sequence at a guanosine (G) because transcription of the native U6 snRNA initiates at a G. Note the following:

- If G is part of the target sequence, then incorporate the G into the stem sequence in the top-strand oligo and add a complementary C to the 3' end of the top-strand oligo.
- If G is not the first base of the target sequence, we recommend adding a G to the 5' end of the top-strand oligo directly following the CACC overhang sequence. In this case, do not add the complementary C to the 3' end of the top-strand oligo. Note: We have found that adding the complementary C in this situation can result in reduced activity of the shRNA. Alternative, if use of a G to initiate transcription is not desired, use an adenosine (A) rather than C or T. Note, however, that use of any nucleotide other than G may affect initiation efficiency and position.

Citations & References (5)

Citations & References
Abstract
Structure and function in rhodopsin: a tetracycline-inducible system in stable mammalian cell lines for high-level expression of opsin mutants.
Authors: Reeves Philip J; Kim Jong-Myoung; Khorana H Gobind;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12370422
'Tetracycline-inducible HEK293S stable cell lines have been prepared that express high levels (up to 10 mg/liter) of WT opsin and its mutants only in response to the addition of tetracycline and sodium butyrate. The cell lines were prepared by stable transfection of HEK293S-TetR cells with expression plasmids that contained the ... More
Dual-tagging system for the affinity purification of mammalian protein complexes.
Authors:Giannone RJ, McDonald WH, Hurst GB, Huang Y, Wu J, Liu Y, Wang Y,
Journal:Biotechniques
PubMed ID:17907572
'Although affinity purification coupled with mass spectrometry (MS) provides a powerful tool to study protein-protein interactions, this strategy has encountered numerous difficulties when adapted to mammalian cells. Here we describe a Gateway-compatible dual-tag affinity purification system that integrates regulatable expression, tetracysteine motifs, and various combinations ofaffinity tags to facilitate the ... More
Ectopic expression of necdin induces differentiation of mouse neuroblastoma cells.
Authors: Kobayashi Masakatsu; Taniura Hideo; Yoshikawa Kazuaki;
Journal:J Biol Chem
PubMed ID:12198120
'Necdin is expressed predominantly in postmitotic neurons, and ectopic expression of this protein strongly suppresses cell growth. Necdin has been implicated in the pathogenesis of Prader-Willi syndrome, a human neurodevelopmental disorder associated with genomic imprinting. Here we demonstrate that ectopic expression of necdin induces a neuronal phenotype in neuroblastoma cells. ... More
Cell Cycle Regulation and p53 Activation by Protein Phosphatase 2Calpha.
Authors:Ofek P, Ben-Meir D, Kariv-Inbal Z, Oren M, Lavi S,
Journal:J Biol Chem
PubMed ID:12514180
Protein phosphatase 2C (PP2C) dephosphorylates a broad range of substrates, regulating stress response and growth-related pathways in both prokaryotes and eukaryotes. We now demonstrate that PP2Calpha, a major mammalian isoform, inhibits cell growth and activates the p53 pathway. In 293 cell clones, in which PP2Calpha expression is regulated by a ... More
The gamma -secretase-cleaved C-terminal fragment of amyloid precursor protein mediates signaling to the nucleus.
Authors: Gao Y; Pimplikar S W;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11742091
Sequential processing of the amyloid precursor protein (APP) by beta- and gamma-secretases generates the Abeta peptide, a major constituent of the senile plaques observed in Alzheimer's disease. The cleavage by gamma-secretase also results in the cytoplasmic release of a 59- or 57-residue-long C-terminal fragment (Cgamma). This processing resembles regulated intramembrane ... More