Vybrant™ CFDA SE Cell Tracer Kit
Invitrogen™

Vybrant™ CFDA SE Cell Tracer Kit

Vybrant™ CFDA SE Cell Tracer Kit is used in fluorescent analyses as a long-term tracer of cells. This method isRead more
Have Questions?
Catalog NumberQuantity
V128831 Kit
Catalog number V12883
Price (USD)
305.65
Online exclusive
408.00
Save 102.35
1 kit
Quantity:
1 Kit
Price (USD)
305.65
Online exclusive
408.00
Save 102.35
1 kit
Vybrant™ CFDA SE Cell Tracer Kit is used in fluorescent analyses as a long-term tracer of cells. This method is commonly used for in vitro and in vivo labeling of cells to determine whether or not a cell is proliferating.

• Superior performance—allows for bright, uniform staining of cells
• Long-term retention—dye is well-retained in cells for several days post stain
• Non-toxic—minimal cytotoxicity for in vivo and in vitro assays
• Simple, robust staining protocol

Superior Fluorescent Staining
Successful tracing of cells requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence. The intense fluorescent staining provided by Vybrant™ CFDA SE Cell Tracer Kit enables the visualization of labeled cells for several days post stain, making it an ideal stain for cell tracing studies based on dye dilution.

Long-Term Signal Retention
Vybrant™ CFDA SE dye easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye is designed to provide a consistent signal, even after several days in a cell culture environment.

Non-Toxic
Vybrant™ CFDA SE dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. Researchers have used CFDA SE labeling to show that transplantable hematopoietic cells proliferate in vitro in response to stimulation by a growth factor cocktail.

Simple, Robust Staining Protocol
The Vybrant™ CFDA SE Cell Tracer Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye, thereby extending the shelf life of the dye not dissolved in solution. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1 μl of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeCFDA SE (CFSE)
Excitation Wavelength Range492⁄517
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
FormSolid
FormatTube(s), Slide(s)
Product LineVybrant
Quantity1 Kit
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Label TypeOther Labels or Dyes
Product TypeTracer
Unit Size1 kit
Contents & Storage
Contains 10 vials of 5(6)-CFDA, SE (CFSE, 500 μg per vial), and 1 vial of DMSO (1.3 mL). Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

I'm trying to label my cells in suspension and track them over time by labeling with CFDA SE, but after labeling them they will no longer adhere to a surface (whereas unlabeled cells adhere well). I've tried to label at 10 and 20 µM. What is causing this?

This dye will bind to proteins via primary amines. Too high of a concentration can lead to cell toxicity or unintended modification of cell function. The solution is to reduce the concentration and/or staining time, or to go with a non-protein-binding tracking reagent, such as Qtracker cell labeling reagents.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to do a cell migration study for around 4 hours and need to fluorescently label the cells with a dye. What do you recommend?

Calcein, AM and FDA (fluorescein diaceate) are examples of some dyes used for this application. Since these dyes are not incorporated or covalently attached to any cellular components, they may have a short retention time as some cell types may actively efflux the dye out of the cells. The CellTracker and CellTrace dyes include either a mild thiol-reactive chloromethyl group or amine-reactive succinnimidyl ester group to allow for covalent binding to cellular components, providing for better retention. As with any reagent, one should empirically determine retention times for the cell type used.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I labeled my cells with Calcein, AM, but when I imaged the next day, there was no fluorescence from Calcein. Why?

Calcein, AM is a good choice for cell tracking and as a general cytoplasmic stain. However, it doesn't bind to anything and may be actively pumped out of the cells within a couple hours, which is likely what happened. The retention of Calcein within live cells is dependent upon the inherent properties of the cell type and culture conditions.

For long-term imaging, you may wish to consider a reactive cytoplasmic stains such as CFDA, SE or the CellTracker and CellTrace dyes.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to track my cells with a nucleic acid stain, like DAPI or Hoechst dye. Do you recommend this?

This is not recommended. When these stains bind to DNA and RNA, they may affect the normal function of the nucleic acids, disrupting transcription, as well as replication. Other reagents, such as CellTracker dyes or Qtracker reagents are more optimized for tracking without disrupting normal activity. If a nuclear label is still desired, though, and the cells are mammalian and non-hematopoietic, CellLight nuclear reagents can transiently transfect cells to express GFP or RFP on a nuclear-expressing protein for up to several days without affecting function.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

I want to track my cells over time, and you have a lot of options to choose from. How do I pick the right one?

Please see this Web link (http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-tracing-tracking-and-morphology/cell-tracking.html) to help you choose the right option for your application. Start by planning how long you want to track your cells, then consider the mechanism of binding. Calcein dyes are very uniform in label and are good for short-term cell migration, but may be rapidly effluxed from some cell types. Lipophilic cyanine dyes, such as DiI, DiO, and similar dyes label cell membranes, don’t disrupt function, and can last longer, but have the potential to cross to other cells if membranes fuse. They are also lost upon permeabilization. CellTracker dyes are better for longer-term labeling, as they possess a mildly reactive chloromethyl moiety that allows covalent binding to cellular components. CFDA SE also covalently binds to cellular components. With all the reagents, their retention within cells is dependent upon the rate of cell division and the inherent properties of the cell (active efflux, membrane and protein turnover rates, etc.) and reagents that allow for covalent attachment exhibit longer retention than those that do not.

The longest-lasting and brightest options are the Qtracker reagents, which are taken up through endocytosis. These are so bright individual quantum dots can be detected, and are also robust enough to survive not only fixation and permeabilization, but even the heat and solvents used in paraffin processing.

Find additional tips, troubleshooting help, and resources within our Cell Tracing and Tracking Support Center.

Citations & References (477)

Citations & References
Abstract
Fas/Fas ligand interactions promote activation-induced cell death of NK T lymphocytes.
Authors:Leite-de-Moraes MC,Herbelin A,Gouarin C,Koezuka Y,Schneider E,Dy M
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:11035073
Phenotypic and functional effects of heat shock protein 90 inhibition on dendritic cell.
Authors:Bae J,Mitsiades C,Tai YT,Bertheau R,Shammas M,Batchu RB,Li C,Catley L,Prabhala R,Anderson KC,Munshi NC
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:17548610
Calcein: a novel marker for lymphocytes which enter lymph nodes.
Authors:Weston SA, Parish CR
Journal:Cytometry
PubMed ID:1451604
Previous studies have identified unique cell surface antigens which are associated with the specific binding of lymphocytes to high endothelial venules (HEV). Evidence is presented in this paper which demonstrates that uptake of the fluorescent dye calcein by lymphocytes represents an additional marker for the lymph node homing subpopulation of ... More
Primitive, quiescent, Philadelphia-positive stem cells from patients with chronic myeloid leukemia are insensitive to STI571 in vitro.
Authors:Graham SM, Jørgensen HG, Allan E, Pearson C, Alcorn MJ, Richmond L, Holyoake TL
Journal:Blood
PubMed ID:11756187
In clinical trials, the tyrosine kinase inhibitor STI571 has proven highly effective in reducing leukemic cell burden in chronic myeloid leukemia (CML). The overall sensitivity of CML CD34(+) progenitor cells to STI571 and the degree to which cell death was dependent on cell cycle status were determined. Stem cells (Lin(-)CD34(+)) ... More
Platelet-endothelial interaction in tumor angiogenesis and microcirculation.
Authors:Manegold PC, Hutter J, Pahernik SA, Messmer K, Dellian M
Journal:Blood
PubMed ID:12584142
Activated platelets release angiogenic growth factors and have therefore been proposed to contribute to tumor angiogenesis within a potentially prothrombotic tumor microcirculation. The aim of the study was to investigate interactions of platelets with the angiogenic microvascular endothelium of highly vascularized solid tumors during growth and in response to endothelial ... More