Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining
Actual product may vary
Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining
Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining
Invitrogen™

Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining

Easily distinguish cells undergoing apoptosis with Vybrant Apoptosis Assay Kits, which offer dyes, including YO-PRO-1 and Hoechst 33342, for apoptotic and necrotic cell staining.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeDescription
V13244Hoechst 33342, Propidium IodideVybrant Apoptosis Assay Kit #5
V13243YO-PRO-1, propidium iodideVybrant Apoptosis Assay Kit #4
V23200Alexa Fluor 350 Streptavidin, Propidium Iodide, Biotin-X Annexin VVybrant Apoptosis Assay Kit #6
Catalog number V13244
Price (USD)
268.65
Online exclusive
292.00
Save 23.35
200 assays
Label or Dye:
Hoechst 33342, Propidium Iodide
Description:
Vybrant Apoptosis Assay Kit #5
Price (USD)
268.65
Online exclusive
292.00
Save 23.35
200 assays
Apoptotic cells undergo several physiological processes, including changes in cell membrane permeability and chromatin condensation, which can be distinguished using different dyes in apoptotic and necrotic cell staining protocols. Vybrant Apoptosis Assay kits offer streptavidin biotin-annexin V, propidium iodide (PI), YO-PRO-1, and Hoechst 33342 dyes to differentiate live from apoptotic or necrotic cells, which can then be visualized using flow cytometry.
Vybrant Apoptosis Assay kits offer quick and convenient cell staining dye-based methodologies for differentiating apoptotic and necrotic versus healthy cells, which can then be visualized using flow cytometry.

The Vybrant Apoptosis Assay Kit #4 detects apoptosis via changes that occur in cell membrane permeability. This kit contains ready-to-use solutions of both YO-PRO-1 and propidium iodide (PI) nucleic acid stains. YO-PRO-1 stain selectively passes through the plasma membranes of apoptotic cells and labels them with moderate green fluorescence. Necrotic cells are stained fluorescent red with PI.

The Vybrant Apoptosis Assay Kit #5, or the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry, provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. This kit contains ready-to-use solutions of blue-fluorescent Hoechst 33342 dye, which stains the condensed chromatin of apoptotic cells more brightly than the chromatin of non-apoptotic cells, and red-fluorescent PI dye, which stains dead cells.

The Vybrant Apoptosis Assay Kit #6 provides a rapid and convenient assay for apoptosis, using the bright, blue-fluorescent Alexa Fluor 350 conjugate of streptavidin in conjunction with biotin-X annexin V to detect apoptotic cell populations either by flow cytometry or imaging. In addition, the kit includes a ready-to-use solution of the fluorescent red PI nucleic acid–binding dye. PI is impermeant to live cells and apoptotic cells, but stains dead cells with red fluorescence, binding tightly to their nucleic acid. The kit contains an ample amount of each reagent for about 50 flow cytometric assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue, Red
DescriptionVybrant Apoptosis Assay Kit #5
Excitation/EmissionHoechst: 350/461
propidium iodide: 535/617
Flow Cytometer Laser Lines488, UV
For Use With (Equipment)Flow Cytometer
Label TypeOther Label(s) or Dye(s)
Label or DyeHoechst 33342, Propidium Iodide
Product LineVybrant
Product TypeApoptosis Kit
Quantity200 assays
Shipping ConditionRoom Temperature
Detection MethodFluorescence
FormatTube
Unit Size200 assays
Contents & Storage
1 vial of Hoechst 33342 and 1 vial of propidium iodide. Store in refrigerator (2–8°C) and protect from light

Frequently asked questions (FAQs)

What are the fluorescence excitation/emission maxima for the dyes contained in the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry aka Vybrant Apoptosis Assay Kit #5, Hoechst 33342/Propidium Iodide)?

Hoechst 33342: 350/461 nm, bound to DNA
Propidium iodide: 535/617 nm, bound to DNA

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What kinds of cell health and viability assays can be performed by flow cytometry?

The following cell health and viability assays can be performed by flow cytometry :

-Apoptosis Assays:
Membrane Asymmetry: Annexin V is a member of a family of structurally related proteins that bind phospholipids in the presence of Ca2+. Annexin V binds several phospholipids, but shows highest affinity for phosphatidylserine.
Phosphatidylserine is normally found in the inner leaflet of the cell membrane; however, in the early stages of apoptosis, phosphatidylserine is observed to translocate to the outer leaflet. This translocation makes phosphatidylserine available for annexin V binding in the presence of Ca2+ containing incubation buffer. Cells undergoing apoptosis will stain with annexin V, while normal cells will not. annexin V is available conjugated with a wide range of fluorophores.

Mitochondrial Health: A distinctive feature of the early stages of apoptosis is the disruption of the mitochondria, including changes in membrane and redox potential. We exclusively offer a number of fluorescent probes for analyzing mitochondrial activity in live cells by flow cytometry, with minimal disruption of cellular function.

The MitoProbe family of mitochondrial stains (MitoProbe DiOC2(3) Assay Kit, Cat. No. M34150, MitoProbe JC-1 Assay Kit, Cat. No. M34152, and MitoProbe DiIC1(5) Assay Kit, Cat. No. M34151) provides quick, easy, and reliable flow cytometric detection of the loss of mitochondrial membrane potential that occurs during apoptosis.

Caspase Activity: The CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427) enables flow cytometric detection of activated caspase-3 and caspase-7 in apoptotic cells. The kit includes the novel fluorogenic substrate CellEvent Caspase-3/7 Green Detection Reagent which targets the recognition sequence for activated caspase-3 and caspase-7, as well as SYTOX AADvanced Dead Cell Stain.

DNA Fragmentation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. DNA fragmentation that occurs during apoptosis produces DNA strand breaks, and can be analyzed using TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) assays. The APO-BrdU TUNEL assay (Cat. No. A23210) is a two-color assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by imaging or flow cytometry.

Nuclear Chromatin Condensation: The later stages of apoptosis are characterized by changes in nuclear morphology, including DNA fragmentation, chromatin condensation, degradation of nuclear envelope, nuclear blebbing, and DNA strand breaks. Cells undergoing apoptosis display an increase in nuclear chromatin condensation. As the chromatin condenses, cell-permeable nucleic acid stains becomes hyperfluorescent, thus enabling the identification of apoptotic cells when combined with a traditional dead-cell stain. The Vybrant Apoptosis Assay Kit #5, Hoechst 33342/Propidium Iodide (Cat. No. V13244) provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. The Chromatin Condensation & Membrane Permeability Dead Cell Apoptosis Kit with Hoechst 33342, YO-PRO-1, and PI dyes, for flow cytometry (Cat. No. V23201) detects apoptotic cells with changes in nuclear chromatin condensation and plasma membrane permeability.

-Cell Cycle Analysis:
Live cell assays: The Vybrant DyeCycle family of dyes offers robust fluorescent dyes for live-cell cycle analysis with limited cytotoxicity using 405 nm (Cat. No. V35003), 488 nm (Cat. No. V35004), 532 nm (Cat. No. V35005), or 633 nm (Cat. Nos. V10309 and V10273) excitation. The dyes have low cytotoxicity, allowing stained cells to be sorted and otherwise cultured or assessed with functional assays after staining.

Fixed cell assays: Analyzing cell cycle using FxCycle Violet Stain (Cat. No. F10347), SYTOX AADvanced Dead Cell Stain Kit (Cat. No. S10349) or FxCycle Far Red Stain (Cat. No. F10348) allows for multiple color options for simplified fixed cell cycle analysis.

-Cell Proliferation:
Dye dilution assays for cell proliferation: Dye dilution assays for cell proliferation rely on cell membrane–permeant fluorescent molecules. Upon entry into the cell, the dye will covalently bind to amine groups on proteins, resulting in long-term dye retention within the cell. Through subsequent cell divisions, each daughter cell receives approximately half the fluorescence of the parent. Analysis of the fluorescence intensities of cell populations by flow cytometry enables determination of the number of generations through which a cell or population has progressed since the label was applied. CellTrace fluorescent stains can be used without affecting morphology or physiology to trace generations in vivo or in vitro. There is no known effect on proliferative ability or biology of cells and they are well retained in cells for several days post-stain. Available kits for flow cytometry include CellTrace CFSE Cell Proliferation Kit (Cat. No. C34554), CellTrace Violet Cell Proliferation Kit (Cat. No. C34557), and CellTrace Far Red Cell Proliferation Kit (Cat. No. C34564).

DNA Synthesis Assays: Measuring the synthesis of new DNA is a precise way to assay cell proliferation in individual cells or in cell populations. DNA synthesis–based cell proliferation assays measure the rate of new DNA synthesis based on incorporation of modified nucleosides. The Click-iT Plus EdU cell proliferation assay utilizes the power of click chemistry and the modified nucleoside EdU to provide a superior alternative to BrdU staining for detecting and quantitating newly synthesized DNA. The Click-iT Plus EdU cell proliferation assay is available with Pacific Blue (Cat. No. C10636), Alexa Fluor 488 (Cat. Nos. C10632 and C10633), and Alexa Fluor 647 (Cat. Nos. C10634 and C10635).

-Viability Assays:
Dead cells often give false positive results, as they tend to bind non-specifically to many reagents. Therefore, removing dead cells from your flow cytometry data is a critical step to help ensure accurate results and analysis.

Non-fixable Membrane Permeability Stains: SYTOX Dead Cell Stains (Cat. Nos. S34857, S34860, S34861, S34859, and S34862) do not cross intact cell membranes, and they exhibit increased fluorescence upon dsDNA binding, making them some of our most brilliant dead cell stains. Cell-impermeant classic DNA-binding dyes include propidium iodide (Cat. No. P21493) and 7-AAD (Cat. No. A1310). Both of these dyes have been used extensively for viability assays in flow cytometry. CellTrace Calcein AM dyes can be passively loaded into adherent and nonadherent cells. These cell-permeant esterase substrates serve as viability probes that measure both enzymatic activity, which is required to activate their fluorescence, and cell membrane integrity, which is required for intracellular retention of their fluorescent products. Available with blue (Cat. No. C34853), violet (Cat. No. C34858), and green (Cat. No. C34852) fluorescence, these dyes are ideal for short-term staining of live cells and can be used in multiplexed flow cytometry experiments.

Fixable Viability Stains: The LIVE/DEAD Fixable Dead Cell Stains are fixable viability dyes that help to ensure accurate assessment of cell viability in samples after fixation and/or permeabilization. LIVE/DEAD Fixable Dead Cell Stain Kits are based on the reaction of a fluorescent reactive dye with cellular proteins (amines). These dyes cannot penetrate live-cell membranes, so only cell-surface proteins are available to react with the dye, resulting in dim staining. The reactive dye can permeate the damaged membranes of dead cells and stain both the interior and exterior amines, resulting in more intense staining. LIVE/DEAD Fixable Dead Cell Stain Kits are available in eight single-channel colors available for UV, 405, 488, 532, 561, or 633 nm lasers in three packaging sizes to match your experiment.



Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

DAPI and Hoechst dyes are quite similar to each other. Why would I choose one over the other?

DAPI is a very common blue-fluorescent dye for nuclear counterstaining and gives very bright labeling on nuclei in fixed and permeabilized cells and tissues. However, it is considered to be a semi-permeant to impermeant stain and provides inconsistent staining of live cells. Hoechst 33342 dye is cell-permeant and stains with the same binding mechanism and fluorescent color; it is preferred for live-cell imaging and is just as good as DAPI for fixed cell labeling.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (3)

Citations & References
Abstract
Stimulation of CD95 (Fas) blocks T lymphocyte calcium channels through sphingomyelinase and sphingolipids.
Authors:Lepple-Wienhues A, Belka C, Laun T, Jekle A, Walter B, Wieland U, Welz M, Heil L, Kun J, Busch G, Weller M, Bamberg M, Gulbins E, Lang F
Journal:Proc Natl Acad Sci U S A
PubMed ID:10570152
'Calcium influx through store-operated calcium release-activated calcium channels (CRAC) is required for T cell activation, cytokine synthesis, and proliferation. The CD95 (Apo-1/Fas) receptor plays a role in self-tolerance and tumor immune escape, and it mediates apoptosis in activated T cells. In this paper we show that CD95-stimulation blocks CRAC and ... More
Mechanism of neurodegeneration of neurons with mitochondrial DNA mutations.
Authors:Abramov AY, Smulders-Srinivasan TK, Kirby DM, Acin-Perez R, Enriquez JA, Lightowlers RN, Duchen MR, Turnbull DM,
Journal:Brain
PubMed ID:20157008
Mutations of mitochondrial DNA are associated with a wide spectrum of disorders, primarily affecting the central nervous system and muscle function. The specific consequences of mitochondrial DNA mutations for neuronal pathophysiology are not understood. In order to explore the impact of mitochondrial mutations on neuronal biochemistry and physiology, we have ... More
Pit cells (Hepatic natural killer cells) of the rat induce apoptosis in colon carcinoma cells by the perforin/granzyme pathway.
Authors:Vermijlen D, Luo D, Robaye B, Seynaeve C, Baekeland M, Wisse E
Journal:Hepatology
PubMed ID:9862849
The high mortality of colon cancer is to a large extent caused by the frequent occurrence of liver metastasis. This is remarkable, because the liver harbors two distinct cell populations that can eliminate invading cancer cells, namely hepatic natural killer (NK) cells and Kupffer cells. These hepatic NK cells, known ... More