Tampón de unión de anexina (5X), para citometría de flujo
Tampón de unión de anexina (5X), para citometría de flujo
Have Questions?
Número de catálogo
Cantidad
V13246
50 mL
Número de catálogo V13246
Precio (MXN)
-
Cantidad:
50 mL
This product is designed to facilitate the binding of annexin V to phosphatidylserine for use in apoptosis assays.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (aplicación)Citometría de flujo
Formularioconcentrado
Formulación50 mm de HEPES, 700 mm de NaCl, 12,5 mm de CaCl2, pH 7,4
Tipo de productoTampón de unión de anexina
Cantidad50 mL
Condiciones de envíoHielo húmedo
pH7,4
Unit Size50 mL
Contenido y almacenamiento
Contiene 1 frascos de tampón de unión de anexina (solución de 5x, 50 ml).
Almacenar de 2–8 °C.
Preguntas frecuentes
I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?
Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).
Can I detect annexin V staining in an imaging assay?
Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.
When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?
Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).
Yes, this is possible. We have established protocols for annexin V staining combined with intracellular staining of lymphocytes that can be found here. The most important step is to leave some binding buffer in the suspension when fixation is started. Compared to staining of live cells, the intensity of the annexin V signal may be somewhat reduced.
Citations & References (6)
Citations & References
Abstract
Surface functionalization of doxorubicin-loaded liposomes with octa-arginine for enhanced anticancer activity.
Doxorubicin-loaded PEGylated liposomes (commercially available as DOXIL or Lipodox) were surface functionalized with a cell-penetrating peptide, octa-arginine (R8). For this purpose, R8-peptide was conjugated to the polyethylene glycol-dioleoyl phosphatidylethanolamine (PEG-DOPE) amphiphilic co-polymer. The resultant R8-PEG-PE conjugate was introduced into the lipid bilayer of liposomes at 2 mol% of total lipid ... More
Bruton's tyrosine kinase is at the crossroads of metabolic adaptation in primary malignant human lymphocytes.
Authors:Sharif-Askari B, Doyon D, Paliouras M, Aloyz R
Journal:Sci Rep
PubMed ID:31363127
'In this work we explored metabolic aspects of human primary leukemic lymphocytes that hold a potential impact on the treatment of Bruton tyrosine kinase (BTK)-driven diseases. Our results suggest that there is crosstalk between Bruton tyrosine kinase (BTK) signaling and bioenergetic stress responses. In primary chronic lymphocytic leukemia (CLL) lymphocytes, ... More
Ibrutinib Resistance Is Reduced by an Inhibitor of Fatty Acid Oxidation in Primary CLL Lymphocytes.
Authors:Galicia-Vázquez G, Aloyz R
Journal:Front Oncol
PubMed ID:30319974
Chronic Lymphocytic Leukemia (CLL) is an incurable disease, characterized by the accumulation of malignant B-lymphocytes in the blood stream (quiescent state) and homing tissues (where they can proliferate). In CLL, the targeting of B-cell receptor signaling through a Burton's tyrosine kinase inhibitor (ibrutinib) has rendered outstanding clinical results. However, complete ... More
A tyrosine kinase-activating variant Asn666Ser in PDGFRB causes a progeria-like condition in the severe end of Penttinen syndrome.
Authors:Bredrup C, Stokowy T, McGaughran J, Lee S, Sapkota D, Cristea I, Xu L, Tveit KS, Høvding G, Steen VM, Rødahl E, Bruland O, Houge G
Journal:Eur J Hum Genet
PubMed ID:30573803
Missense variants located to the "molecular brake" in the tyrosine kinase hinge region of platelet-derived growth factor receptor-ß, encoded by PFGFRB, can cause Penttinen-type (Val665Ala) and Penttinen-like (Asn666His) premature ageing syndromes, as well as infantile myofibromatosis (Asn666Lys and Pro660Thr). We have found the same de novo PDGFRB c.1997A>G p.(Asn666Ser) variants ... More
Functional Autophagic Flux Regulates AgNP Uptake And The Internalized Nanoparticles Determine Tumor Cell Fate By Temporally Regulating Flux.
Authors:Fageria L, Bambroo V, Mathew A, Mukherjee S, Chowdhury R, Pande S
Journal:Int J Nanomedicine
PubMed ID:31819419
Silver nanoparticles (AgNPs) are known to induce the conserved, cellular, homeostatic process- autophagy in tumor cells. Previous studies primarily focus on the pro-survival role of autophagy post AgNP exposure in tumor cells, but seldom on its role in AgNP uptake, or on the functional significance of autophagy temporal dynamics. Our ... More