Product Image
Invitrogen™

pPICZ A, B, & C Pichia Vectors

pPICZ A、B、&C Pichiaベクターは、組換えタンパク質の簡単なクローニングと選択、高レベルでの発現、および迅速な検出と浄化のために設計されています。これらのベクターには、複数コピー一体型株を直接選択するためのゼオシン™耐性遺伝子が含まれています詳細を見る
製品番号(カタログ番号)数量
V1902020 μg
製品番号(カタログ番号) V19020
価格(JPY)
107,500
キャンペーン価格
Ends: 25-Sep-2026
153,600
割引額 46,100 (30%)
20 µg
数量:
20 μg
pPICZ A、B、&C Pichiaベクターは、組換えタンパク質の簡単なクローニングと選択、高レベルでの発現、および迅速な検出と浄化のために設計されています。これらのベクターには、複数コピー一体型株を直接選択するためのゼオシン™耐性遺伝子が含まれています。ゼオシン™の量を増やして選択することで、ゲノムに統合された目的の遺伝子の複数コピーがある株が得られます。組換えPichia株で目的遺伝子のコピー数を増やすと、発現レベルが高くなる可能性があります。このベクターはEasySelect™ Pichia Expression Kit(カタログ番号K1740-01)に含まれています。

pPICZベクターの特長は以下のとおりです。

•抗myc抗体での便利な検出のためのPichia pastoris
c-
mycエピトープタグによる高レベルでの発現用の誘導型AOX1プロモーター
• ニッケルキレートレジンによる迅速な精製用のC末端ポリヒスチジン(6xHis)タグと抗His(C-term)抗体による検出
• 複数コピー一体型株を直接選択するためのZeocin™耐性遺伝子

研究用にのみ使用できます。診断用には使用いただけません。
仕様
抗生物質耐性菌ゼオシン™(ZeoR)
製品タイプPichia発現ベクター
数量20 μg
ベクターpPIC
クローニング法制限酵素/MCS
プロモーターAOX1
タンパク質タグHisタグ(6x)、c-Mycエピトープタグ, c-Myc Epitope Tag
Unit Size20 µg
組成および保存条件
10 mMのTris – HClで懸濁された1 mMのEDTA、pH 8.0のそれぞれ20 µgのpPICZ A、B、&C。GS115/pPICZ/lacZポジティブコントロール株も含まれています。

このベクターは-5~-30℃で保管してください。

よくあるご質問(FAQ)

When selecting for blasticidin-resistant transformants in the X-33 strain using pPIC6/pPIC6α vectors, why do I get large and small colonies on YPD plates containing 300 µg/ml blasticidin?

Generally, large colonies represent transformants containing pPIC6/pPIC6α integrants, while small colonies represent transformants containing pPIC6/pPIC6α non-integrants. These non-integrants have transduced the pPIC6/pPIC6α plasmid, and therefore, exhibit a low level of blasticidin resistance in the initial selection process. Upon subsequent screening, these non-integrant transformants do not retain blasticidin resistance.

When choosing a blasticidin-resistant transformant for your expression studies, we recommend that you pick blasticidin-resistant colonies from the initial transformation plate and streak them on a second YPD plate containing the appropriate concentration of blasticidin. Select transformants that remain blasticidin-resistant for further studies.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My transformation is not working. Do you have any suggestions?

Here are some suggestinos:

- Make sure that you have harvested cells during log-phase growth (OD <1.0 generally).
- If electroporation is being used, see the electroporator manual for suggested conditions. Vary electroporation parameters if necessary.
- Use more DNA.
- Use freshly made competent cells.
- If the LiCl transformation method is being used, try boiling the carrier DNA.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

My spheroplasting of Pichia worked twice, but hasn't worked since. The OD of the culture simply does not drop.

Here are some things to consider:

- If the OD of cells that are used is too high, they will not spheroplast. Do not overgrow cells.
- Do not use old cells and make sure that they are in log phase of growth.
- Make sure to mix zymolyase well before using. Zymolyase is more of a suspension than a solution.
- Make the PEG solution fresh each time and check the pH.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

What are the different kinds of media used for culturing Pichia pastoris and S. cerevisiae?

Following are the rich and minimal media used for culturing Pichia pastoris and S. cerevisiae:

Rich Media:
S. cerevisiae and Pichia pastoris
YPD (YEPD): yeast extract, peptone, and dextrose
YPDS: yeast extract, peptone, dextrose, and sorbitol

Pichia pastoris only
BMGY: buffered glycerol-complex medium
BMMY: buffered methanol-complex medium

Minimal Media (also known as drop-out media):
S. cerevisiae
SC (SD): Synthetic complete (YNB, dextrose (or raffinose or galactose), and amino acids)

Pichia pastoris
MGY: minimal glycerol medium
MD: minimal dextrose
MM: minimal methanol
BMGH: buffered minimal glycerol
BMMH: buffered minimal methanol

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Is there a recommended protocol for fermentation using constitutive expression vectors such as pGAPZ?

Use the following high cell density protocol for pGAP clones. Feed carbon until the desired density is reached (300 to 400 g/L wet cell weight (WCW)). If the protein is well-behaved in the fermenter, increase to 300-400 g/L WCW as with methanol inducible clones. These densities can be reached in less than 48 hours of fermentation. We have fermented constitutive expressers on glycerol using these protocols with good results. Some modifications to the Fermentation Basal Salts Medium that you might want to make are:

1) Substitute 2% dextrose for the 4% glycerol in the batch medium.
2) Substitute 40% dextrose for the 50% glycerol in the fed-batch medium.
3) Feed the 40% dextrose at 12 mL/L/hr (Jim Cregg has published data on expression using several carbon sources as substrates; dextrose gave the highest levels of expression).
4) Yeast extract and peptone may be added to the medium for protein stability.

One warning: If you are working with His- strains, they remain His- after transformation with pGAPZ. Fermentation in minimal medium will require addition of histidine to the fermenter.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

引用および参考文献 (3)

引用および参考文献
Abstract
In vivo functional assay of a recombinant aquaporin in Pichia pastoris.
Authors:Daniels MJ, Wood MR, Yeager M,
Journal:Appl Environ Microbiol
PubMed ID:16461705
The water channel protein PvTIP3;1 (alpha-TIP) is a member of the major intrinsic protein (MIP) membrane channel family. We overexpressed this eukaryotic aquaporin in the methylotrophic yeast Pichia pastoris, and immunogold labeling of cellular cryosections showed that the protein accumulated in the plasma membrane, as well as vacuolar and other ... More
The crystal structure of human CD21: Implications for Epstein-Barr virus and C3d binding.
Authors: Prota Andrea E; Sage David R; Stehle Thilo; Fingeroth Joyce D;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12122212
Human complement receptor type 2 (CD21) is the cellular receptor for Epstein-Barr virus (EBV), a human tumor virus. The N-terminal two short consensus repeats (SCR1-SCR2) of the receptor interact with the EBV glycoprotein gp350/220 and also with the natural CD21 ligand C3d. Here we present the crystal structure of the ... More
The donor substrate specificity of the human beta 1,3-glucuronosyltransferase I toward UDP-glucuronic acid is determined by two crucial histidine and arginine residues.
Authors: Ouzzine Mohamed; Gulberti Sandrine; Levoin Nicolas; Netter Patrick; Magdalou Jacques; Fournel-Gigleux Sylvie;
Journal:J Biol Chem
PubMed ID:11986319
The human beta1,3-glucuronosyltransferase I (GlcAT-I) plays a key role in proteoglycan biosynthesis by catalyzing the transfer of glucuronic acid onto the trisaccharide-protein linkage structure Galbeta1,3Galbeta1,4Xylbeta-O-Ser, a prerequisite step for polymerization of glycosaminoglycan chains. In this study, we identified His(308) and Arg(277) residues as essential determinants for the donor substrate (UDP-glucuronic ... More