Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining
Invitrogen™

Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining

Easily distinguish cells undergoing apoptosis with Vybrant Apoptosis Assay Kits, which offer dyes, including YO-PRO-1 and Hoechst 33342, for apoptotic and necrotic cell staining.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeDescription
V23200Alexa Fluor 350 Streptavidin, Propidium Iodide, Biotin-X Annexin VVybrant Apoptosis Assay Kit #6
V13243YO-PRO-1, propidium iodideVybrant Apoptosis Assay Kit #4
V13244Hoechst 33342, Propidium IodideVybrant Apoptosis Assay Kit #5
Catalog number V23200
Price (USD)
372.65
Online exclusive
497.00
Save 124.35
50 assays
Label or Dye:
Alexa Fluor 350 Streptavidin, Propidium Iodide, Biotin-X Annexin V
Description:
Vybrant Apoptosis Assay Kit #6
Price (USD)
372.65
Online exclusive
497.00
Save 124.35
50 assays
Apoptotic cells undergo several physiological processes, including changes in cell membrane permeability and chromatin condensation, which can be distinguished using different dyes in apoptotic and necrotic cell staining protocols. Vybrant Apoptosis Assay kits offer streptavidin biotin-annexin V, propidium iodide (PI), YO-PRO-1, and Hoechst 33342 dyes to differentiate live from apoptotic or necrotic cells, which can then be visualized using flow cytometry.
Vybrant Apoptosis Assay kits offer quick and convenient cell staining dye-based methodologies for differentiating apoptotic and necrotic versus healthy cells, which can then be visualized using flow cytometry.

The Vybrant Apoptosis Assay Kit #4 detects apoptosis via changes that occur in cell membrane permeability. This kit contains ready-to-use solutions of both YO-PRO-1 and propidium iodide (PI) nucleic acid stains. YO-PRO-1 stain selectively passes through the plasma membranes of apoptotic cells and labels them with moderate green fluorescence. Necrotic cells are stained fluorescent red with PI.

The Vybrant Apoptosis Assay Kit #5, or the Chromatin Condensation/Dead Cell Apoptosis Kit with Hoechst 33342 and PI for Flow Cytometry, provides a rapid and convenient assay for apoptosis based on fluorescence detection of the compacted state of the chromatin in apoptotic cells. This kit contains ready-to-use solutions of blue-fluorescent Hoechst 33342 dye, which stains the condensed chromatin of apoptotic cells more brightly than the chromatin of non-apoptotic cells, and red-fluorescent PI dye, which stains dead cells.

The Vybrant Apoptosis Assay Kit #6 provides a rapid and convenient assay for apoptosis, using the bright, blue-fluorescent Alexa Fluor 350 conjugate of streptavidin in conjunction with biotin-X annexin V to detect apoptotic cell populations either by flow cytometry or imaging. In addition, the kit includes a ready-to-use solution of the fluorescent red PI nucleic acid–binding dye. PI is impermeant to live cells and apoptotic cells, but stains dead cells with red fluorescence, binding tightly to their nucleic acid. The kit contains an ample amount of each reagent for about 50 flow cytometric assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorBlue, Red
DescriptionVybrant Apoptosis Assay Kit #6
Excitation/EmissionAlexa Fluor™ 346/442: 346
PI: 535/617
Flow Cytometer Laser LinesUV, 488
For Use With (Equipment)Flow Cytometer
Label TypeAlexa Fluor™ Dyes, Biotin & Other Haptens, Other Label(s) or Dye(s)
Label or DyeAlexa Fluor 350 Streptavidin, Propidium Iodide, Biotin-X Annexin V
Product LineAlexa Fluor
Product TypeApoptosis Kit
Quantity50 assays
Shipping ConditionWet Ice
Detection MethodFluorescence
FormatTube
Unit Size50 assays
Contents & Storage
1 vial of biotin-x annexin V, 1 vial of Alexa Fluor™ 350 streptavidin, 1 vial of propidium iodide, and 1 bottle of 5x annexin binding buffer. Store in refrigerator (2–8°C) and protect from light

Citations & References (5)

Citations & References
Abstract
mTOR-dependent proliferation defect in human ES-derived neural stem cells affected by myotonic dystrophy type 1.
Authors:Denis JA, Gauthier M, Rachdi L, Aubert S, Giraud-Triboult K, Poydenot P, Benchoua A, Champon B, Maury Y, Baldeschi C, Scharfmann R, Piétu G, Peschanski M, Martinat C,
Journal:J Cell Sci
PubMed ID:23444380
'Patients with myotonic dystrophy type 1 exhibit a diversity of symptoms that affect many different organs. Among these are cognitive dysfunctions, the origin of which has remained elusive, partly because of the difficulty in accessing neural cells. Here, we have taken advantage of pluripotent stem cell lines derived from embryos ... More
Glucagon-like peptide 1 stimulates post-translational activation of glucokinase in pancreatic beta cells.
Authors:Ding SY, Nkobena A, Kraft CA, Markwardt ML, Rizzo MA,
Journal:J Biol Chem
PubMed ID:21454584
Glucagon-like peptide 1 (GLP-1) potentiates glucose-stimulated insulin secretion from pancreatic ß cells, yet does not directly stimulate secretion. The mechanisms underlying this phenomenon are incompletely understood. Here, we report that GLP-1 augments glucose-dependent rises in NAD(P)H autofluorescence in both ßTC3 insulinoma cells and islets in a manner consistent with post-translational ... More
Nuclear expression of E2F4 induces cell death via multiple pathways in normal human intestinal epithelial crypt cells but not in colon cancer cells.
Authors:Garneau H, Alvarez L, Paquin MC, Lussier C, Rancourt C, Tremblay E, Beaulieu JF, Rivard N,
Journal:Am J Physiol Gastrointest Liver Physiol
PubMed ID:17656449
E2F transcription factors control cell cycle progression. The localization of E2F4 in intestinal epithelial cells is cell cycle dependent, being cytoplasmic in quiescent differentiated cells but nuclear in proliferative cells. However, whether nuclear translocation of E2F4 alone is sufficient to trigger intestinal epithelial cell proliferation remains to be established. Adenoviruses ... More
G protein-coupled receptor-promoted trafficking of Gbeta1gamma2 leads to AKT activation at endosomes via a mechanism mediated by Gbeta1gamma2-Rab11a interaction.
Authors:García-Regalado A, Guzmán-Hernández ML, Ramírez-Rangel I, Robles-Molina E, Balla T, Vázquez-Prado J, Reyes-Cruz G,
Journal:Mol Biol Cell
PubMed ID:18701709
G-protein coupled receptors activate heterotrimeric G proteins at the plasma membrane in which most of their effectors are intrinsically located or transiently associated as the external signal is being transduced. This paradigm has been extended to the intracellular compartments by studies in yeast showing that trafficking of Galpha activates phosphatidylinositol ... More
Tetramethylpyrazine regulates breast cancer cell viability, migration, invasion and apoptosis by affecting the activity of Akt and caspase-3.
Authors:Shen J, Zeng L, Pan L, Yuan S, Wu M, Kong X
Journal:Oncol Lett
PubMed ID:29541225
Tetramethylpyrazine (TMP), an effective component of the traditional Chinese medicine Chuanxiong Hort, has been proven to exhibit a beneficial effect in a number of types of malignant epithelial cancer. However, the mode of action of TMP on breast cancer cells remains unknown. The aim of the present study was to ... More