Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry
Invitrogen™

Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry

Assess cell viability with Dead Cell Apoptosis kits with Annexin V and conjugated fluorescent dyes such as Alexa Fluor 488, FITC, PI, PE, APC, and SYTOX Green.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityLabel or Dye
V351131 KitSYTOX Green, APC
V1324150 AssaysAlexa Fluor 488, Propidium Iodide
V132421 KitFITC, Propidium Iodide
V351121 KitSYTOX Green, R-PE
V13245250 AssaysAlexa Fluor 488, Propidium Iodide
Catalog number V35113
Price (USD)
754.00
1 kit
Quantity:
1 Kit
Label or Dye:
SYTOX Green, APC
Price (USD)
754.00
1 kit
Easily differentiate live, dead, and apoptotic cells during flow cytometry with our Dead Cell Apoptosis kits with Annexin V conjugates including Alexa Fluor 488, FITC, propidium iodide, PE, APC, and SYTOX Green. These conjugates distinguish live, dead, or apoptotic cells via different dyes, which is vital for confirming cell viability and apoptosis using multi-parametric studies.
There are several advantages to using Dead Cell Apoptosis kits with Annexin V for flow cytometry in order to assay cell viability:

Superior brightness
Unlike other Annexin V kits that have lower protein concentrations or purification levels, the Annexin V Alexa Fluor 488 conjugate is optimized for flow cytometry to provide the largest separation between apoptotic and live cells. The Alexa Fluor 488 dye is a superior green-fluorescent dye with a spectrum similar to fluorescein (FITC).

High binding efficiency
Annexin V conjugates are made from a highly purified cys-annexin, resulting in higher binding efficiency and extremely accurate characterization of the apoptotic process.

Multi-parametric
Many publications require a minimum of two different ways to identify apoptotic cells. This multi-parametric kit detects phosphatidyl serine (PS) on the cytoplasmic surface of the cell membrane, while membrane integrity is determined via propidium iodide.

Dead Cell Apoptosis Kit with Annexin V Alexa Fluor 488 & PI
The Dead Cell Apoptosis Kit with Annexin V Alexa Fluor 488 & Propidium Iodide (PI) (V13241, V13245) is used in flow cytometry to measure early apoptosis by detecting phosphatidyl serine (PS) expression and membrane permeability. When cells are stained with Annexin V and propidium iodide, apoptotic cells expressing PS show green fluorescence, which can be detected in the FITC channel, and low red fluorescence. Dead or necrotic cells show bright red fluorescence and no green fluorescence, while live cells show no green or red fluorescence.

Dead Cell Apoptosis Kit with Annexin V PE and SYTOX Green
The Dead Cell Apoptosis Kit with Annexin V PE and SYTOX Green detects PS externalization in apoptotic cells during flow cytometry using recombinant annexin V conjugated to the orange fluorescent phycobiliprotein R-PE, while dead cells are detected with SYTOX Green nucleic acid stain. After treatment with both probes, apoptotic cells show orange fluorescence, dead cells show green fluorescence, and live cells show little or no fluorescence. These populations can easily be distinguished in the 530/30 nm and 585/42 nm bandpass filters with a 488 nm laser flow cytometer.

Dead Cell Apoptosis Kit with Annexin V APC and SYTOX Green
The Dead Cell Apoptosis Kit with Annexin V APC and SYTOX Green detects PS externalization in apoptotic cells during flow cytometry using recombinant annexin V conjugated to red laser-excited allophycocyanin, and dead cells using SYTOX Green nucleic acid stain. Apoptotic cells are detected by annexin V binding to externalized PS. Live cells show little or no fluorescence, apoptotic cells show red fluorescence and very little green fluorescence, and late apoptotic cells show a higher level of red and orange fluorescence. These populations can easily be distinguished using a flow cytometer that has both 488 nm and 633 nm excitation sources (an argon-ion laser and a HeNe laser).

The Dead Cell Apoptosis Kit with Annexin V FITC and PI
The Dead Cell Apoptosis Kit with Annexin V FITC and PI detects PS externalization in apoptotic cells using recombinant annexin V conjugated to green-fluorescent FITC dye and dead cells using PI. Necrotic cells that stain with PI show red fluorescence. After treatment with both probes, apoptotic cells show green fluorescence, dead cells show red and green fluorescence, and live cells show little or no fluorescence.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDead Cell Apoptosis Kit with Annexin V APC and SYTOX Green, for flow cytometry
Excitation/Emission503, 650/524, 660
Flow Cytometer Laser Lines633/635, 488
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Label or DyeSYTOX Green, APC
No. of Reactions50
Product TypeApoptosis Detection Kit
Quantity1 Kit
Shipping ConditionWet Ice
Unit Size1 kit
Contents & Storage
Contains 1 vial of annexin V, APC conjugate (250 μL), 1 vial of SYTOX green stain (100 μL),and 1 bottle of annexin binding buffer (5X solution, 15 mL). Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

How do SYTO dyes bind to DNA?

The binding mode of SYTO nucleic acid stains is unknown. However, the behavior of these and related nucleic acid dyes suggests the following binding properties:

1.They appear to contact the solvent (suggested by sensitivity to salt, divalent cations, and in particular, SDS) and thus are likely to have contacts in the grooves.
2.All SYTO dyes appear to show some base selectivity and are thus likely to have minor groove contacts.
3.They can be removed from nucleic acid via ethanol precipitation; this characteristic is not shared by ethidium bromide and other intercalators. Likewise, the dyes are not removed from nucleic acid via butanol or chloroform extraction. These extraction methods do remove ethidium bromide from nucleic acid. 4. SYTO binding is not affected by nonionic detergents.
5. SYTO dyes are not quenched by BrdU, so they do not bind nucleic acids in precisely the same way as Hoechst 33342 and DAPI ((4′,6-diamidino-2-phenylindole).

SYBR Green I has shown little mutagenicity on frameshift indicator strains, indicating that it isn't likely to strongly intercalate.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When using Dead Cell Apoptosis Kits with Annexin V for Flow Cytometry, what does a PI+ only population (hence Annexin V negative) correspond to?

These products are cells that died without undergoing apoptosis. A PI+ only population could be either a result of experimental condition or sample preparation.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (4)

Citations & References
Abstract
Preclinical Evaluation of AMG 925, a FLT3/CDK4 Dual Kinase Inhibitor for Treating Acute Myeloid Leukemia.
Authors:Keegan K, Li C, Li Z, Ma J, Ragains M, Coberly S, Hollenback D, Eksterowicz J, Liang L, Weidner M, Huard J, Wang X, Alba G, Orf J, Lo MC, Zhao S, Ngo R, Chen A, Liu L, Carlson T, Quéva C, McGee LR, Medina J, Kamb A, Wickramasinghe D, Dai K,
Journal:
PubMed ID:24526162
'Acute myeloid leukemia (AML) remains a serious unmet medical need. Despite high remission rates with chemotherapy standard-of-care treatment, the disease eventually relapses in a major proportion of patients. Activating Fms-like tyrosine kinase 3 (FLT3) mutations are found in approximately 30% of patients with AML. Targeting FLT3 receptor tyrosine kinase has ... More
Temozolomide-mediated DNA methylation in human myeloid precursor cells: differential involvement of intrinsic and extrinsic apoptotic pathways.
Authors:Wang H, Cai S, Ernstberger A, Bailey BJ, Wang MZ, Cai W, Goebel WS, Czader MB, Crean C, Suvannasankha A, Shokolenkoc I, Wilson GL, Baluyut AR, Mayo LD, Pollok KE,
Journal:Clin Cancer Res
PubMed ID:23536437
'An understanding of how hematopoietic cells respond to therapy that causes myelosuppression will help develop approaches to prevent this potentially life-threatening toxicity. The goal of this study was to determine how human myeloid precursor cells respond to temozolomide (TMZ)-induced DNA damage. We developed an ex vivo primary human myeloid precursor ... More
Peiminine serves as an adriamycin chemosensitizer in gastric cancer by modulating the EGFR/FAK pathway.
Authors:Tang Q, Wang Y, Ma L, Ding M, Li T, Nie Y, Gu Z
Journal:Oncol Rep
PubMed ID:29328433
Gastric cancer (GC) is one of the most common malignancies of the digestive tract. Adriamycin (ADR) has been widely utilized in various chemotherapy regimens for treating GC, yet its long-term application may increase drug resistance resulting in treatment failure. Increasing evidence shows that bioactive natural products can be used as ... More
LncRNA HOTTIP Participates in Cisplatin Resistance of Tumor Cells by Regulating miR-137 Expression in Pancreatic Cancer.
Authors:Yin F, Zhang Q, Dong Z, Hu J, Ma Z
Journal:Onco Targets Ther
PubMed ID:32280243
This study aimed to investigate the effect of HOTTIP and miR-137 on cisplatin resistance of pancreatic cancer cells, and study the mechanism of the effect of HOTTIP on the resistance to cisplatin in pancreatic cancer cells, so as to provide new targets for clinical treatment of pancreatic cancer. ... More