Product Image
Invitrogen™
pLenti6.2/V5-DEST™ Gateway™ Vector
The pLenti6.2⁄V5-DEST™ Gateway™ Vector is a Gateway™-adapted ViraPower™ II lentiviral expression vector for lentiviral-based expression of a target gene inLeia mais
Have Questions?
Número do catálogoQuantity
V368206 μg
Número do catálogo V36820
Preço (BRL)
-
Quantity:
6 μg
The pLenti6.2⁄V5-DEST™ Gateway™ Vector is a Gateway™-adapted ViraPower™ II lentiviral expression vector for lentiviral-based expression of a target gene in dividing and non-dividing mammalian cells. The vector has the CMV promoter for driving constitutive expression of the target gene and the PGK promoter for driving long-term, persistent expression of the Blasticidin stable selection marker.
Advantages
• Stable expression
• Long-term experiments
• High-throughput screening
• Accurate titer of functional virus (using Blasticidin method)
• Flexible and versatile Gateway™ recombination cloning technology
Key Features
• Human cytomegalovirus (CMV) immediate early promoter to control high-level expression of the gene of interest
• PKG Promoter for expression of Blasticidin selection marker
Kit includes
• pLenti6.2⁄V5-DEST™ Gateway™ Vector box
• One Shot™ Stbl3™ chemically competent E. coli (C7373-03)
Related SKUs
• 293FT Cell Line (R70007; R70007)
• ViraPower™ Lentiviral Support Kit (K4970-00)
• ViraPower™ Lentiviral Packaging Mix (K4975-00)
• Lipofectamine™ 2000 (11668-019; 11668-027)
For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Constitutive or Inducible SystemConstitutive
Delivery TypeLentiviral
For Use With (Application)Viral Expression
Product TypeLentiviral Expression Vector
Quantity6 μg
Selection Agent (Eukaryotic)Blasticidin
Selection Marker PromoterPGK Promoter
VectorpLenti
Cloning MethodGateway
Product LineGateway, ViraPower
PromoterCMV
Protein TagV5 Epitope Tag
Unit Size6 µg
Conteúdo e armazenamento
pLenti6.2⁄V5-DEST™ Gateway™ Vector Box contains pLenti6.2⁄V5-DEST, 40 μl of 150 ng⁄μl vector in 10 mM Tris-HCl, 1mM EDTA, pH 8.0 and pLenti6.2⁄V5-GW⁄lacZ control, 20 μl of 0.5 μg⁄μl vector in 10 mM Tris-HCl, 1mM EDTA, pH 8.0. Store at –20°C.
One Shot™ Stbl3™ chemically competent E. coli box contains transformation reagents including 20 single-use 50 μl aliquots of One Shot™ Competent Cells, S.O.C. medium, and pUC19 supercoiled plasmid control. Store at -80°C.
Frequently asked questions (FAQs)
Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?
In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.
Do you have a recommended single-step protocol for BP/LR recombination?
Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.
How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?
We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.
Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?
We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.
Do you offer Gateway vectors for expression in plants?
We do not offer any Gateway vectors for expression in plants.