Product Image
Invitrogen™

pcDNA™3.1/His A, B, & C Mammalian Expression Vectors

All pcDNA™ vectors contain a strong promoter for high-level expression in mammalian cells, a choice of selection marker for generatingRead more
Have Questions?
Catalog NumberQuantity
V38520
also known as V385-20
20 μg
Catalog number V38520
also known as V385-20
Price (TWD)
44,610.00
Online offer
Ends: 31-Dec-2026
63,715.00
Save 19,105.00 (30%)
20 µg
Quantity:
20 μg
Price (TWD)
44,610.00
Online offer
Ends: 31-Dec-2026
63,715.00
Save 19,105.00 (30%)
20 µg
All pcDNA™ vectors contain a strong promoter for high-level expression in mammalian cells, a choice of selection marker for generating stable cell lines, and an epitope tag for easy detection with a monoclonal antibody and rapid purification on nickel-chelating resin. Each vector is available in three reading frames to simplify cloning in-frame with the fusion tag.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CleavageEK (Enterokinase) Recognition Site
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Constitutive Expression
Product TypeMammalian Expression Vector
Quantity20 μg
Selection Agent (Eukaryotic)Geneticin™ (G-418)
VectorpcDNA
Cloning MethodRestriction Enzyme/MCS
Product LinepcDNA
PromoterCMV
Protein TagHis Tag (6x), Xpress Epitope Tag
Unit Size20 µg
Contents & Storage

Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

What is the difference between pcDNA3.1 vectors and the pcDNA3.3-TOPO vector?

pcDNA3.1 vectors contain the core CMV promoter that is truncated before the start of transcription, whereas the pcDNA 3.3-TOPO vector has the 672 bp native CMV promoter. This native CMV promoter allows high-level gene expression with two- to five-fold higher protein yields compared to other expression vectors. pcDNA3.1 vectors are available in restriction, TOPO, and Gateway cloning versions and as untagged and epitope-tagged versions, whereas the pcDNA3.3-TOPO vector is a TOPO TA-adapted, untagged vector that can be used to express native proteins without extraneous amino acids, and is hence ideal for antibody production and structural biology.

Citations & References (54)

Citations & References
Abstract
LMP-1, a LIM-domain protein, mediates BMP-6 effects on bone formation.
Authors:Boden SD, Liu Y, Hair GA, Helms JA, Hu D, Racine M, Nanes MS, Titus L
Journal:Endocrinology
PubMed ID:9832452
Glucocorticoids can promote osteoblast differentiation from fetal calvarial cells and bone marrow stromal cells. We recently reported that glucocorticoid specifically induced bone morphogenetic protein-6 (BMP-6), a glycoprotein signaling molecule that is a multifunctional regulator of vertebrate development. In the present study, we used fetal rat secondary calvarial cultures to determine ... More
DBC2, a candidate for a tumor suppressor gene involved in breast cancer.
Authors:Hamaguchi Masaaki; Meth Jennifer L; von Klitzing Christine; Wei Wen; Esposito Diane; Rodgers Linda; Walsh Tom; Welcsh Piri; King Mary-Claire; Wigler Michael H;
Journal:Proc Natl Acad Sci U S A
PubMed ID:12370419
A previously uncharacterized gene, DBC2 (deleted in breast cancer), was cloned from a homozygously deleted region at human chromosome 8p21. DBC2 contains a highly conserved RAS domain and two putative protein interacting domains. Our analyses indicate that DBC2 is the best candidate tumor suppressor gene from this region. It lies ... More
Characterization and channel coupling of the P2Y(12) nucleotide receptor of brain capillary endothelial cells.
Authors:Simon Joseph; Filippov Alexander K; Göransson Sara; Wong Yung H; Frelin Christian; Michel Anton D; Brown David A; Barnard Eric A;
Journal:J Biol Chem
PubMed ID:12080041
Rat brain capillary endothelial (B10) cells express an unidentified nucleotide receptor linked to adenylyl cyclase inhibition. We show that this receptor in B10 cells is identical in sequence to the P2Y(12) ADP receptor ( ... More
Rab11-FIP2 functions in transferrin recycling and associates with endosomal membranes via its COOH-terminal domain.
Authors: Lindsay Andrew J; McCaffrey Mary W;
Journal:J Biol Chem
PubMed ID:11994279
'Rab11-FIP2 is a recently described member of the Rip11/Rab11-FIP/Rab coupling protein family of Rab11 interacting proteins. Rab11-FIP2 interacts with both Rab11 and myosin Vb and co-localizes with Rab11 in both HeLa and Madin-Darby canine kidney cells (Hales, C. M., Griner, R., Hobdy-Henderson, K. C., Dorn, M. C., Hardy, D., Kumar, ... More
N-terminal processing is essential for release of epithin, a mouse type II membrane serine protease.
Authors: Cho E G; Kim M G; Kim C; Kim S R; Seong I S; Chung C; Schwartz R H; Park D;
Journal:J Biol Chem
PubMed ID:11567025
'Epithin was originally identified as a mouse type II membrane serine protease. Its human orthologue membrane type-serine protease 1 (MT-SP1)/matriptase has been reported to be localized on the plasma membrane. In addition, soluble forms of matriptase were isolated from human breast milk and breast cancer cell-conditioned medium. In this paper, ... More