Product Image
Invitrogen™

pBAD/gIII Kit

The pBAD/gIII Kit provides all of the necessary reagents to express recombinant proteins under the tight regulation of the araBADRead more
Have Questions?
Catalog NumberQuantity
V450011 kit
Catalog number V45001
Price (USD)
1,278.00
1 kit
Quantity:
1 kit
Price (USD)
1,278.00
1 kit
The pBAD/gIII Kit provides all of the necessary reagents to express recombinant proteins under the tight regulation of the araBAD promoter. To simplify purification, the vector pBAD/gIII encodes a leader peptide which directs the recombinant protein into the
periplasmic space. The vector provides:

• The araBAD promoter for tightly regulated expression
• Translation initiation signals for optimized E. coliexpression
• Leader peptide from the bacteriophage fd gene III protein (gIII) for secreted expression
• C-terminal polyhistidine (6xHis) tag for purification with nickel-chelating resin
• C-terminal c-myc epitope for detection and analysis with an Anti-myc Antibody

A set of three vectors is provided (A, B, and C). Each has the C-terminal tag in a different reading frame relative to the multiple cloning site to simplify in-frame cloning of your gene.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Constitutive or Inducible SystemInducible
Inducing AgentArabinose
Product TypeBacterial Expression Vector
Quantity1 kit
Selection Agent (Eukaryotic)None
VectorpBAD
Cloning MethodRestriction Enzyme/MCS
PromoteraraBAD
Protein Tagc-Myc Epitope Tag
Unit Size1 kit
Contents & Storage
The pBAD/gIII Kit includes 20 μg each of pBAD/gIII A, B, & C, 20 μg of pBAD/gIII/calmodulin, 1 ml 20% L-arabinose, LMG194 and TOP10 E. coli stabs. Store stabs at 2 - 8°C. Store all other components at -20°C. All components are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

How much L-arabinose should I use to induce expression with the pBAD expression system?

While the amount of L-arabinose can vary depending on your expression experiment, we suggest performing a pilot expression experiment with varying amounts of L-arabinose from 0.00002% to 0.2%.

Should I use TOP10 cells or the LMG194 E. coli strain you offer for expression with my pBAD system?

Top10

Advantages:
- Saves time, can go directly from cloning to expression.
- The glycerol stock is more stable because these strains are endA- and recA-.

Disdvantages:
- This strain is not protease-deficient. Therefore, the protein may be degraded.

LMG194

Advantages:
- Grows well in minimal media, except M9.
-Have to transform the plasmid into the cells just for expression.
-RM medium with glucose to ensure low basal level of protein.

Disadvantages:
- Not protease-deficient. Therefore, the protein may be degraded.
- The glycerol stock may not be stable because this cell strain is not recA- or endA-.

What competent cells do you recommend I use for expression with my pBAD expression system?

We recommend using a competent cell strain that is araBADC- and araEFGH+, allowing transportation of L-arabinose, but not metabolizing it. This is important for expression studies, as the level of L-arabinose will be constant inside the cell and will not decrease over time. We offer our TOP10 competent cells, or our LMG194 E. coli strain.

Can you tell me the sequence of primers that can be used in pBAD vectors to sequence my gene of interest?

The pBAD vectors contain a forward and reverse pBAD primer flanking the gene of interest. The sequences are as follows:

pBAD forward primer: 5'-ATGCCATAGCATTTTTATCC-3'

pBAD reverse primer: 5'-GATTTAATCTGTATCAGG-3'

Do you have a list of the cap colors for the pBAD vectors?

Here are the cap colors:

- pBAD/His A: Red

- pBAD/His B: Orange

- pBAD/His C: Yellow

- pBAD/His LacZ: Green

- pBad/gIII A: Yellow

- pBad/gIII B: Green

- pBad/gIII C: Blue

- pBAD/gIII/calmodulin: Purple

Citations & References (4)

Citations & References
Abstract
Inhibition of adipocyte differentiation by resistin-like molecule alpha. Biochemical characterization of its oligomeric nature.
Authors: Blagoev Blagoy; Kratchmarova Irina; Nielsen Mogens M; Fernandez Minerva M; Voldby Jesper; Andersen Jens S; Kristiansen Karsten; Pandey Akhilesh; Mann Matthias;
Journal:J Biol Chem
PubMed ID:12189153
A novel family of cysteine-rich secreted proteins with unique tissue distribution has recently been identified. One of the members, resistin (for
The COOH Terminus of Arylamine N-Acetyltransferase from Salmonella typhimurium Controls Enzymic Activity.
Authors: Mushtaq Adeel; Payton Mark; Sim Edith;
Journal:J Biol Chem
PubMed ID:11799105
Arylamine N-acetyltransferases (NATs) are a homologous family of enzymes, which acetylate arylamines, arylhydroxylamines, and arylhydrazines by acetyl transfer from acetyl-coenzyme A (Ac-CoA) and are found in many organisms. NAT was first identified as the enzyme responsible for the inactivation of the anti-tubercular drug isoniazid in humans. The three-dimensional structure of ... More
Specificity determinants and diversification of the Brassica self-incompatibility pollen ligand.
Authors:Chookajorn T, Kachroo A, Ripoll DR, Clark AG, Nasrallah JB,
Journal:Proc Natl Acad Sci U S A
PubMed ID:14694189
Self-incompatibility in crucifers is effected by allele-specific interactions between the highly polymorphic stigmatic S locus receptor kinase (SRK) and its pollen ligand, the S locus cysteine-rich protein (SCR). Here we show that specificity in SCR function is determined by four contiguous amino acids in one variant, indicating that the minimum ... More
A general method for greatly improving the affinity of antibodies by using combinatorial libraries.
Authors:Rajpal A, Beyaz N, Haber L, Cappuccilli G, Yee H, Bhatt RR, Takeuchi T, Lerner RA, Crea R,
Journal:Proc Natl Acad Sci U S A
PubMed ID:15939870
Look-through mutagenesis (LTM) is a multidimensional mutagenesis method that simultaneously assesses and optimizes combinatorial mutations of selected amino acids. The process focuses on a precise distribution within one or more complementarity determining region (CDR) domains and explores the synergistic contribution of amino acid side-chain chemistry. LTM was applied to an ... More