The pAd⁄CMV⁄V5-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ adenoviral expression vector, pAd⁄CMV⁄V5-DEST™ vector for easy recombination-based cloning and adenoviral-based,Read more
Have Questions?
Catalog Number
Quantity
V49320
also known as V493-20
6 μg
Catalog number V49320
also known as V493-20
Price (USD)
1,782.00
6 µg
Quantity:
6 μg
Price (USD)
1,782.00
6 µg
The pAd⁄CMV⁄V5-DEST™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ adenoviral expression vector, pAd⁄CMV⁄V5-DEST™ vector for easy recombination-based cloning and adenoviral-based, transient expression of a target gene in dividing and non-dividing mammalian cells. The vector allows generation of an adenovirus containing the target gene where constitutive, high-level expression is driven by the CMV promoter.
Advantages • High efficiency and rapid recombination cloning • Produces high titer adenoviral stocks • Efficient delivery of the gene to dividing and non-dividing mammalian cells in vitro or in vivo • Produces replication-incompetent virus for enhanced biosafety of the system • Amenable for use in high-throughput applications
Key Features • Gateway™ Technology for efficient and rapid cloning • CMV promoter for high-level constitutive expression of gene of interest • Human Ad5 sequences (ΔE3) and Viral Inverted Terminal Repeats (ITRs) for packaging of the expression construct into virions • Herpes Simplex Virus thymidine kinase (TK) polyadenylation sequence for efficient transcription termination and polyadenylation of mRNA • V5 epitope for detection of recombinant protein • Ampicillin selection marker
Kit Includes • pAd⁄CMV⁄V5-DEST™ Gateway™ Vector • pAd⁄CMV⁄V5-GW⁄lacZ control plasmid
For research use only. Not intended for any therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeAdenoviral
For Use With (Application)Viral Expression
Product TypeMammalian Expression Vector
Quantity6 μg
VectorpDEST
Cloning MethodGateway
Product LineGateway, ViraPower
PromoterCMV
Protein TagV5 Epitope Tag
Unit Size6 µg
Contents & Storage
• pAd-CMV⁄V5-DEST™ Vector 150 ng⁄μl in TEBuffer, pH 8.0. 40 μl (Store at -20°C) • pAd⁄CMV⁄V5-GW⁄lacZ control plasmid 1 μg⁄μl in TE Buffer, pH 8.0. 10 μl (Store at -20°C)
Frequently asked questions (FAQs)
Can I perform the single-step protocol for the BP/LR Clonase reaction using BP Clonase enzyme and LR Clonase enzyme instead of BP Clonase II enzyme and LR Clonase II enzyme?
In the single-step protocol for the BP/LR Clonase reaction, we would not recommend substituting the BP Clonase II/LR Clonase II enzymes with BP Clonase /LR Clonase enzymes as this would result in very low recombination efficiency.
Do you have a recommended single-step protocol for BP/LR recombination?
Yes, we have come up with a single-step protocol for BP/LR Clonase reaction (http://www.thermofisher.com/us/en/home/life-science/cloning/gateway-cloning.html#1), where DNA fragments can be cloned into Destination vectors in a single step reaction, allowing you to save time and money.
How can I move my gene of interest from a Gateway-adapted expression clone to a new Destination vector as I have lost the entry clone?
We would recommend performing a BP reaction with a Donor vector in order to obtain an entry clone. This entry clone can then be used in an LR reaction with the Destination vector to obtain the new expression clone.
Can I purchase the 5X LR Clonase buffer or 5X BP Clonase buffer separately?
We do not offer the 5X LR Clonase buffer and 5X BP Clonase buffer as standalone products. They are available as part of the enzyme kits.
Do you offer Gateway vectors for expression in plants?
We do not offer any Gateway vectors for expression in plants.
Disruption of glucose sensing and insulin secretion by ribozyme Kir6.2-gene targeting in insulin-secreting cells.
Authors:Li L, Rojas A, Wu J, Jiang C,
Journal:Endocrinology
PubMed ID:15166124
'The ATP-sensitive K+ (KATP) channel, composed of Kir6.2 and sulfonylurea receptor (SUR1), in pancreatic beta-cells is believed to serve as a metabolic sensor regulating insulin secretion according to glucose levels. Thus, genetic disruption of Kir6.2 expression may impair KATP channel function in glucose sensing and insulin secretion. Here we show ... More
The receptor for parathyroid hormone and parathyroid hormone-related peptide is hydrolyzed and its signaling properties are altered by directly binding the calpain small subunit.
Authors:Shimada M, Mahon MJ, Greer PA, Segre GV,
Journal:Endocrinology
PubMed ID:15691895
'We show calcium-dependent, direct binding between the N-terminal portion of the PTH/PTHrP receptor (PTH1R) C-terminal intracellular tail and the calpain small subunit. Binding requires, but may not be limited to, amino acids W474, S475, and W477. The wild-type, full-length rat (r) PTH1R, but not rPTH1R with W474A/W477A substitutions, copurifies with ... More
Authors:Zhang Y, Ma K, Sadana P, Chowdhury F, Gaillard S, Wang F, McDonnell DP, Unterman TG, Elam MB, Park EA,
Journal:J Biol Chem
PubMed ID:17079227
The pyruvate dehydrogenase complex (PDC) catalyzes the conversion of pyruvate to acetyl-CoA in mitochondria and is a key regulatory enzyme in the oxidation of glucose to acetyl-CoA. Phosphorylation of PDC by the pyruvate dehydrogenase kinases (PDK2 and PDK4) inhibits PDC activity. Expression of the PDK genes is elevated in diabetes ... More
Adipose-specific expression, phosphorylation of Ser794 in insulin receptor substrate-1, and activation in diabetic animals of salt-inducible kinase-2.
Authors:Horike N, Takemori H, Katoh Y, Doi J, Min L, Asano T, Sun XJ, Yamamoto H, Kasayama S, Muraoka M, Nonaka Y, Okamoto M,
Journal:J Biol Chem
PubMed ID:12624099
Salt-inducible kinase (SIK), first cloned from the adrenal glands of rats fed a high salt diet, is a serine/threonine protein kinase belonging to an AMP-activated protein kinase family. Induced in Y1 cells at an early stage of ACTH stimulation, it regulated the initial steps of steroidogenesis. Here we report the ... More