Product Image
Invitrogen™

pcDNA™5/FRT/TO Vector Kit

pcDNA™FRT⁄TO vector is a 5.1 kb inducible expression vector designed for use with the Flp-In™ T-REx™ System. The control plasmid,자세히 알아보기
Have Questions?
카탈로그 번호수량
V652020
V6520-20으로도 사용됨
20 μg
카탈로그 번호 V652020
V6520-20으로도 사용됨
제품 가격(KRW)
1,291,000
온라인 행사
Ends: 31-Mar-2026
1,475,000
할인액 184,000 (12%)
20 µg
수량:
20 μg
제품 가격(KRW)
1,291,000
온라인 행사
Ends: 31-Mar-2026
1,475,000
할인액 184,000 (12%)
20 µg
pcDNA™FRT⁄TO vector is a 5.1 kb inducible expression vector designed for use with the Flp-In™ T-REx™ System. The control plasmid, pcDNA™5⁄FRT⁄TO⁄CAT, is also included for use as a positive control for transfection and expression in your Flp-In™ T-REx™ host cell line of choice.

The pcDNA™5⁄FRT⁄TO vector contains the following elements:

• A hybrid human cytomegalovirus (CMV)⁄TetO2 promoter for high-level, tetracycline-regulated expression of the gene of interest in a wide range of mammalian cells
• Multiple cloning site with 10 unique restriction sites to facilitate cloning the gene of interest
• FLP Recombination Target (FRT) site for Flp recombinase-mediated integration of the vector into the Flp-In™ T-REx™ host cell line
• Hygromycin resistance gene for selection of stable cell lines
For Research Use Only. Not for use in diagnostic procedures.
사양
구성 또는 유도성 시스템Inducible
배달 유형Transfection
용도(애플리케이션)Regulated Expression
유도제Tetracycline
제품 유형Flp-In/T-REx System Expression Vector
수량20 μg
선택 제제(진핵)Hygromycin
벡터pcDNA
클로닝 방법Restriction Enzyme/MCS
제품라인Flp-In, T-REx, pcDNA
프로모터CMV/TO
단백질 태그Untagged
Unit Size20 µg
구성 및 보관
Contains 20 μg of each: pcDNA™FRT⁄TO and pcDNA™FRT⁄TO⁄CAT vectors. Store both vectors at -20°C. All reagents are guaranteed stable for 6 months when properly stored.

자주 묻는 질문(FAQ)

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

Do you offer a GFP-expressing mammalian expression vector that I can use as a control to monitor my transfection and expression?

We offer pJTI R4 Exp CMV EmGFP pA Vector, Cat. No. A14146, which you can use to monitor your transfection and expression.

인용 및 참조 문헌 (2)

인용 및 참조 문헌
Abstract
AMP-activated Kinase Inhibits the Epithelial Na+ Channel through Functional Regulation of the Ubiquitin Ligase Nedd4-2.
Authors:Bhalla V, Oyster NM, Fitch AC, Wijngaarden MA, Neumann D, Schlattner U, Pearce D, Hallows KR,
Journal:J Biol Chem
PubMed ID:16844684
'We recently found that the metabolic sensor AMP-activated kinase (AMPK) inhibits the epithelial Na(+) channel (ENaC) through decreased plasma membrane ENaC expression, an effect requiring the presence of a binding motif in the cytoplasmic tail of the beta-ENaC subunit for the ubiquitin ligase Nedd4-2. To further examine the role of ... More
The MLL fusion gene, MLL-AF4, regulates cyclin-dependent kinase inhibitor CDKN1B (p27kip1) expression.
Authors:Xia ZB, Popovic R, Chen J, Theisler C, Stuart T, Santillan DA, Erfurth F, Diaz MO, Zeleznik-Le NJ,
Journal:Proc Natl Acad Sci U S A
PubMed ID:16169901
MLL, involved in many chromosomal translocations associated with acute myeloid and lymphoid leukemia, has >50 known partner genes with which it is able to form in-frame fusions. Characterizing important downstream target genes of MLL and of MLL fusion proteins may provide rational therapeutic strategies for the treatment of MLL-associated leukemia. ... More