Vanquish™ Split Samplers
Actual product may vary
Vanquish™ Split Samplers
Vanquish™ Split Samplers
Vanquish™ Split Samplers
Thermo Scientific™

Vanquish™ Split Samplers

Gain confidence in peak identification and quantitation from maximum injection precision and huge sample capacity with high-throughput performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBiocompatible VersionIncludes
VC-A13-A-02NoVanquish™ Split Sampler C
VC-A12-A-02NoVanquish™ Split Sampler CT
VF-A10-A-03YesVanquish Split Sampler FT
VH-A10-A-03YesVanquish Split Sampler HT
VH-A14-A-03YesVanquish™ Amplify Split Sampler HT
VF-A14-A-03YesVanquish™ Amplify Split Sampler FT
Catalog number VC-A13-A-02
Price (USD)
-
Biocompatible Version:
No
Includes:
Vanquish™ Split Sampler C

Combine excellent injection precision with huge sample capacity with Thermo Scientific™ Vanquish™ Split Samplers. These samplers employ high-throughput performance to deliver the highest possible confidence in peak identification and quantitation. They provide unsurpassed sample dosing precision and are optimized for ruggedness and uptime, even under system pressures of up to 1500 bar (22,000 psi) and challenging eluent conditions. Now you can record UHPLC data over several days unattended, without running out of samples.

View HPLC/UHPLC resources

Maximum injection precision, huge sample capacity, high-throughput performance
  • SmartInject technology for unrivaled retention time precision and extended column lifetime up to max. 1500bar (22,000 psi)
  • Out-of-the-box vial capacity of 216 samples
  • Increase sample capacity to 23 well plates (up to 8832 samples) by adding the optional Thermo Scientific™ Vanquish™ Charger module
  • By default, all thermostatted Vanquish Samplers utilize air stream cooling to maintain sample integrity
  • Automated sample configuration using barcode reading
  • Transfer of methods is incredibly easy
  • Unlimited application flexibility with customizable gradient delay volume
Ordering Information

Sample capacity: Any four of the following (SBS footprint)

  • 54 x 12 mm OD vials (≤1.5 mL)
  • 96 x 6, 7 and 8 mm OD vials (≤1.2 mL)
  • 16 x 15 mm OD vials (≤4 mL)
  • 9 x 22.5 mm OD vials (≤10 mL)
  • Well plates (96 and 384, deep and shallow)
+ Capacity of 12 x 22.5 mm OD vials (≤10 mL) in the carousel
Specifications
Carryover<0.002% with caffeine
Dimensions (L x W x H)290 mm x 420 mm x 620 mm (11.4 in. x 16.5 in. x 24.4 in.)
IncludesVanquish™ Split Sampler C
Injection Cycle TimeDown to 8 sec. depending on separation conditions
Injection Volume Precision<0.25% area RSD for 3 μL (caffeine in water); Typically <0.5% area RSD for 1 μL (caffeine in water)
PC ConnectionUSB 2.0; 3-port HUB to connect additional Vanquish modules
Temperature RangeSample compartment: not available
Biocompatible VersionNo
Injection TypeSplit Loop
Unit SizeEach