QuEChERS Sodium Sulfate in Sealed Pouches
Thermo Scientific™
QuEChERS Sodium Sulfate in Sealed Pouches
Simplify your sample preparation with Thermo Scientific™ QuEChERS Sodium Sulfate in Sealed Pouches. Ideal for moisture removal in QuEChERS extractions, these pouches ensure consistent and reliable results. Conveniently packed for ease of use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSalt Composition
VD-Q-35 g of Sodium Sulfate
VD-Q-11.5 g of Sodium Sulfate
Catalog number VD-Q-3
Price (USD)
245.00
Each
Salt Composition:
5 g of Sodium Sulfate
Price (USD)
245.00
Each
Thermo Scientific QuEChERS Sodium Sulfate in Sealed Pouches streamline your sample preparation process by effectively removing moisture during QuEChERS extractions. Each pouch is pre-measured for accuracy and convenience, ensuring consistent results across samples. Packaged in a user-friendly format, these pouches are ideal for high-throughput laboratories.
  • Pre-measured sodium sulfate for accurate moisture removal
  • Sealed pouches ensure product integrity and ease of use
  • Ideal for QuEChERS extraction protocols
  • Consistent and reliable results for sample preparation
  • Convenient packaging for high-throughput applications
Specifications
MethodQuEChERS sample preparation
Salt Composition5 g of Sodium Sulfate
Sample Size5 g per sample
Sample TypeAll
Product LineQuechers
SorbentSodium Sulfate
TypeQuEChERS Sample Prep
Volume (Metric)50 mL
Unit SizeEach