Owl™ VEP-3 Large Tank Electroblotting System
Thermo Scientific™

Owl™ VEP-3 Large Tank Electroblotting System

Rapidly transfer high molecular weight proteins with the Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System.
Have Questions?
Catalog NumberType
VEP-3Electroblotting System
Catalog number VEP-3
Price (USD)
2,690.00
Each
Type:
Electroblotting System
Price (USD)
2,690.00
Each
Rapidly transfer high molecular weight proteins with the Thermo Scientific™ Owl™ VEP-3 Large Tank Electroblotting System.
Features:
  • Removable cooling block permits efficient heat transfer.
  • Coded blotting cassettes ensure proper anode/cathode orientation; load easily into buffer chamber.
  • Stir bars (sold separately) may be added to bottom of buffer chamber to circulate buffer.
  • Cassettes allow easy loading of the gel sandwich and locks securely for loading into the buffer chamber.

Includes:

  • Buffer chamber with stainless-steel cathode plate and titanium anode plate
  • Safety interlocking lid with attached power supply leads and cooling module
  • Two blotting cassettes with foam pads
  • 25 sheets of 20W × 18.5cmL (7.88 × 7.25 in.) blotting filter paper
Specifications
Dimensions (L x W x H)3.5 x 11.8 x 30 in. (9 x 30 x 30 cm)
Height (English)30 in.
Height (Metric)30 cm
Length (English)3.5 in.
Length (English) Gel7.8 in.
Length (Metric)9 cm
Length (Metric) Gel20 cm
Volume (English) Buffer1.05 gal.
Volume (Metric) Buffer4 L
Width (English)11.8 in.
Width (English) Gel7.2 in.
Width (Metric)30 cm
Width (Metric) Gel18.5 cm
Gel Size20 x 18.5 cm
Product LineOwl
TypeElectroblotting System
Unit SizeEach