System Bases for Vanquish™
Thermo Scientific™
System Bases for Vanquish™
Add the Vanquish System Base as fundamental part for the installation of the Vanquish HPLC and UHPLC systems in the required setup.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호모델용도(애플리케이션)용도 (장비)최대 압력
VF-S01-A-03System Base Vanquish Horizon and Flex UHPLC SystemsSingle Channel LCVanquish Horizon or Vanquish Flex Modules1517 bar (22,000 psi) or 1034 bar (15,000 psi) depending on configuration
VF-S02-A-03System Base Vanquish Duo UHPLC SystemsSystem Base Vanquish Duo UHPLC SystemsVanquish Duo UHPLC systems700 bar (10,152 psi)
VC-S01-A-03System Base Vanquish Core HPLC SystemsSingle Channel LCVanquish HPLC and UHPLC Systems700 bar (10,152 psi)
VF-S03-A-03System Base Vanquish Core HPLC SystemsSingle Channel LCVanquish HPLC and UHPLC Systems151.7 MPa
카탈로그 번호 VF-S01-A-03
제품 가격(KRW)
-
모델:
System Base Vanquish Horizon and Flex UHPLC Systems
용도(애플리케이션):
Single Channel LC
용도 (장비):
Vanquish Horizon or Vanquish Flex Modules
최대 압력:
1517 bar (22,000 psi) or 1034 bar (15,000 psi) depending on configuration
Add the Thermo Scientific™ Vanquish™ System base to support the installation of Thermo Scientific™ Vanquish™ Core HPLC systems, Thermo Scientific™ Vanquish™ Flex or Horizon UHPLC system Thermo Scientific™ Vanquish™ Duo UHPLC Systems. The system base contains the base module with drawer, a solvent rack, all capillaries required to set up either a single-flow-path or dual-flow path, respectively. The included Thermo Scientific™ Viper™ TQ UHPLC Fittings for confident connections of the fluidic flow paths ease the installation for every user level.
The base and top of your installation
  • The system base is the foundation of the installation and carries all other modules of the LC stack.
  • A drawer in the system base helps to keep your lab clean and tidy by providing a space to store tools and LC accessories.
  • With the system base comes a ship kit, containing all capillaries needed to install standard configurations of the Vanquish HPLC or UHPLC systems.
  • Viper TQ capillaries are provided in biocompatible MP35N for the installation of Vanquish Flex, Horizon and Duo UHPLC systems or in stainless steel material Vanquish Core UHPLC system, respectively, depending on the ordered system base.
  • Viper TQ UHPLC Fittings are torque-controlled Fingertight fittings with audible and tactile feedback for confidence in correct and leak-free installation.
  • The solvent rack provides a secure location for placing the solvent bottles
  • The magnetic front of the solvent rack allows for user-specific system labelling.
사양
용도(애플리케이션)Single Channel LC
모델System Base Vanquish Horizon and Flex UHPLC Systems
제품라인Vanquish Horizon/Flex UHPLC Systems
용도 (장비)Vanquish Horizon or Vanquish Flex Modules
최대 압력1517 bar (22,000 psi) or 1034 bar (15,000 psi) depending on configuration
유형System Base and Solvent Rack
Unit SizeEach