Oxoid™ Vegetable Peptone No. 1
Thermo Scientific™
Oxoid™ Vegetable Peptone No. 1
Thermo Scientific™ Oxoid Vegetable Peptone No. 1 is a meat-free general purpose peptone.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
VG0100K25 kg
VG0100B500 g
0100T
también denominado VG0100T
5 kg
Número de catálogo VG0100K
Precio (MXN)
-
Cantidad:
25 kg
Thermo Scientific™ Oxoid Vegetable Peptone No. 1 is a highly nutritious, meat-free general purpose peptone for growth of bacteria and fungi in culture media or fermentation processes.
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability:
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Para utilizar con (aplicación)Culture media or fermentation processes
Protein SourcePea
Cantidad25 kg
Duración de almacenamiento720 Days from Manufacture
FormularioDehydrated Powder
Tipo de productoPeptone
Unit SizeEach
Contenido y almacenamiento
Dehydrated Powder, Ambient & Dry Storage