Oxoid™ Vegetable Peptone Broth
Thermo Scientific™
Oxoid™ Vegetable Peptone Broth
Thermo Scientific™ Oxoid Vegetable Peptone Broth is a meat-free general-purpose peptone.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
VG0101B500 g
VG0101T5 kg
Número de catálogo VG0101B
Precio (MXN)
-
Cantidad:
500 g
Thermo Scientific™ Oxoid Vegetable Peptone Broth is a highly nutritious, general-purpose peptone.
An exceptional meat-free alternative to Typtone Soya Broth, it delivers equivalent performance in culture media or as a fermentation base.
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability:
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Para uso de laboratorio
Especificaciones
Para utilizar con (aplicación)Culture media or fermentation processes
Protein SourcePea / Yeast
Cantidad500 g
Duración de almacenamiento720 Days from Manufacture
FormularioDehydrated Powder
Tipo de productoPeptone
Unit SizeEach
Contenido y almacenamiento
Dehydrated Powder, Ambient & Dry Storage