Oxoid™ Vegetable Peptone Broth
Oxoid™ Vegetable Peptone Broth
Thermo Scientific™

Oxoid™ Vegetable Peptone Broth

Thermo Scientific™ Oxoid Vegetable Peptone Broth is a meat-free general-purpose peptone.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
VG0101T5 kg
VG0101B500 g
Catalog number VG0101T
Price (BRL)
-
Request A Quote
Quantity:
5 kg
Thermo Scientific™ Oxoid Vegetable Peptone Broth is a highly nutritious, general-purpose peptone.

An exceptional meat-free alternative to Typtone Soya Broth, it delivers equivalent performance in culture media or as a fermentation base.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
For Laboratory Use
Specifications
For Use With (Application)Culture media or fermentation processes
Protein SourcePea / Yeast
Quantity5 kg
Shelf Life720 Days from Manufacture
FormDehydrated Powder
Product TypePeptone
Unit SizeEach
Contents & Storage
Dehydrated Powder, Ambient & Dry Storage