Vanquish™ Column Compartments
Actual product may vary
Vanquish™ Column Compartments
Vanquish™ Column Compartments
Vanquish™ Column Compartments
Thermo Scientific™
Vanquish™ Column Compartments
Control your separation temperature for superior UHPLC performance using Thermo Scientific™ Vanquish™ column compartments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
VH-C10-A-03Vanquish Column Compartment HVanquish Flex UHPLC System, Vanquish Horizon UHPLC System
VC-C10-A-03Vanquish Column Compartment CVanquish Core HPLC System
VN-C10-A-01Vanquish Column Compartment NVanquish Neo UHPLC System
Catalog number VH-C10-A-03
Price (INR)
-
Description:
Vanquish Column Compartment H
For Use With (Equipment):
Vanquish Flex UHPLC System, Vanquish Horizon UHPLC System
Control your separation temperature with confidence using Thermo Scientific™ Vanquish™ Column Compartments. The unique combination of active and/or passive eluent pre-heating and two column thermostatting modes—still and forced air—available in Vanquish Column Compartments H and C, ensures reliable matching of the eluent and column temperature for superior UHPLC performance and allows for easy LC method transfer. Vanquish Column Compartment N supports advanced low-flow applications as well as capillary and micro-flow direct injection applications in the Thermo Scientific™ Vanquish™ Neo UHPLC system.
View HPLC/UHPLC resources
Vanquish Column Compartments H and C offer:
  • Wide column temperature ranges with cooling down to 5 °C (max. 18 K below ambient)
  • Still-air thermostatting for minimum thermal mismatch by viscous heating for best possible efficiencies and resolution under UHPLC conditions
  • Forced-air thermostatting for fast thermal equilibration and highest flexibility, e.g., for method transfer from other instruments
  • Active eluent preheating and post-column cooling (Vanquish Column Compartment H only) for optimum separation control
  • Advanced workflows, such as Tandem LC and LC-MS, online SPE, trap and loop heart-cut 2D-LC with the installation of up to two optional switching valves
Vanquish Column Compartment N offers:
  • Passive pre-heating and forced-air thermostatting (column heating only)
  • Support for advanced low-flow applications as well as capillary and micro-flow direct injection applications with two optional low-dispersion switching valves
  • Optimized system stacking and capillary guiding for MS connection
Specifications
Column Capacity2 x max, 300 mm w/pre-heater or guard column, Max. ID: 10 mm; 2 x max, 250 mm w/pre-heater and guard column, Max. ID: 10 mm
DescriptionVanquish Column Compartment H
For Use With (Equipment)Vanquish Flex UHPLC System, Vanquish Horizon UHPLC System
Product TypeColumn Compartment
Temperature Range5°C (max. 18 K below ambient) to 120°C
Unit SizeEach