Informational:Informational:Ovens are not suitable for use with hazardous vapor. For use with nonflammable, noncorrosive inert gases only.
Thermo Scientific™

Lindberg/Blue M™ Vacuum Ovens

Catalog number: VO1218C
Thermo Scientific™

Lindberg/Blue M™ Vacuum Ovens

Catalog number: VO1218C
Benefit from a digital control, built-in over-temperature protection and a flexible vacuum/purge/release system with Thermo Scientific™ Lindberg/Blue M™ Vacuum Ovens. These ovens work well with a range of applications including drying, curing, outgassing, aging, process control and other applications requiring elevated temperature in reduced atmospheres or vacuum/purge with nonflammable and inert atmospheres.
 
Catalog Number
VO1218C
Unit Size
Each
Description
42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)
42.5 L
Price (USD)
Full specifications
Capacity (English)1.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)42.5 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves3
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
Length (English)26 in.
Length (Metric)66 cm
Width (English)21 in.
Width (English) Interior12 in.
Width (Metric)53 cm
Width (Metric) Interior31 cm
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1250 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
42.5L; 120V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)
42.5 L
Price (USD)
Full specifications
Capacity (English)1.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
DisplayLED
Frequency50/60 Hz
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Length (English)26 in.
Length (Metric)66 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1250 w
Width (English) Interior12 in.
Width (Metric) Interior31 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description42.5L; 120V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)42.5 L
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)21 in.
No. of Shelves3
Width (Metric)53 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
42.5L; 120V; 1250w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)
42.5 L
Price (USD)
Full specifications
Capacity (English)1.5 cu.ft.
Certifications/ComplianceUL listed
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description42.5L; 120V; 1250w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)42.5 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves3
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
Length (English)26 in.
Length (Metric)66 cm
Width (English)21 in.
Width (English) Interior12 in.
Width (Metric)53 cm
Width (Metric) Interior31 cm
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Electrical Requirements120 V 50/60 Hz
Voltage120 V
Wattage1250 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Capacity: 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves
Capacity (Metric)
127.4 L
Price (USD)
Full specifications
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
DisplayLED
Frequency50/60 Hz
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Length (English)24 in.
Length (Metric)61 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1500 w
Width (English) Interior18 in.
Width (Metric) Interior46 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
DescriptionCapacity: 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves
Capacity (Metric)127.4 L
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)27 in.
No. of Shelves2
Width (Metric)69 cm
Warranty1 Year (parts and labor)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
127.4L; 208/240V; 1500w; Painted cabinet; 1 door, 2 shelves
Capacity (Metric)
127.4 L
Price (USD)
Full specifications
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description127.4L; 208/240V; 1500w; Painted cabinet; 1 door, 2 shelves
Capacity (Metric)127.4 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves2
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
Length (English)24 in.
Length (Metric)61 cm
Width (English)27 in.
Width (English) Interior18 in.
Width (Metric)69 cm
Width (Metric) Interior46 cm
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1500 w
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Description
127.4L; 120V; 1500w; SS cabinet; 1 door, 2 shelves
Capacity (Metric)
127.4 L
Price (USD)
Full specifications
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
DisplayLED
Frequency50/60 Hz
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Length (English)24 in.
Length (Metric)61 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1500 w
Width (English) Interior18 in.
Width (Metric) Interior46 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Description127.4L; 120V; 1500w; SS cabinet; 1 door, 2 shelves
Capacity (Metric)127.4 L
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialStainless Steel
Width (English)27 in.
No. of Shelves2
Width (Metric)69 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
18.6L; 120V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)
18.6 L
Price (USD)
Full specifications
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description18.6L; 120V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
18.6L; 208/240V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)
18.6 L
Price (USD)
Full specifications
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage208/240 V
Description18.6L; 208/240V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements208/240 V 50/60 Hz
MaterialPainted Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
18.6L; 120V; 750w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)
18.6 L
Price (USD)
Full specifications
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description18.6L; 120V; 750w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialStainless Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionCapacity (Metric)Price (USD)
VO1218CFull specifications
Each42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves42.5 LRequest A Quote
Capacity (English)1.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description42.5L; 208/240V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)42.5 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves3
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
Length (English)26 in.
Length (Metric)66 cm
Width (English)21 in.
Width (English) Interior12 in.
Width (Metric)53 cm
Width (Metric) Interior31 cm
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1250 w
Frequency50/60 Hz
Unit SizeEach
VO1218AFull specifications
Each42.5L; 120V; 1250w; Painted cabinet; 1 door, 3 shelves42.5 LRequest A Quote
Capacity (English)1.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
DisplayLED
Frequency50/60 Hz
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Length (English)26 in.
Length (Metric)66 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1250 w
Width (English) Interior12 in.
Width (Metric) Interior31 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description42.5L; 120V; 1250w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)42.5 L
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)21 in.
No. of Shelves3
Width (Metric)53 cm
Unit SizeEach
VO1218SAFull specifications
Each42.5L; 120V; 1250w; SS cabinet; 2 door, 3 shelves42.5 LRequest A Quote
Capacity (English)1.5 cu.ft.
Certifications/ComplianceUL listed
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description42.5L; 120V; 1250w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)42.5 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves3
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior18 x 12 x 12 in. (46 x 31 x 31 cm)
Dimensions (L x W x H)26 x 21 x 29 in. (66 x 53 x 74 cm)
Length (English)26 in.
Length (Metric)66 cm
Width (English)21 in.
Width (English) Interior12 in.
Width (Metric)53 cm
Width (Metric) Interior31 cm
Height (English)29 in.
Height (English) Interior12 in.
Height (Metric)74 cm
Height (Metric) Interior31 cm
Depth (English) Interior18 in.
Depth (Metric) Interior46 cm
Shipping Weight (English)250 lb.
Shipping Weight (Metric)113 kg
Electrical Requirements120 V 50/60 Hz
Voltage120 V
Wattage1250 w
Frequency50/60 Hz
Unit SizeEach
VO1824AFull specifications
EachCapacity: 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves127.4 LRequest A Quote
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
DisplayLED
Frequency50/60 Hz
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Length (English)24 in.
Length (Metric)61 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1500 w
Width (English) Interior18 in.
Width (Metric) Interior46 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
DescriptionCapacity: 127.4L; 120V; 1500w; Painted cabinet; One door, Two shelves
Capacity (Metric)127.4 L
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)27 in.
No. of Shelves2
Width (Metric)69 cm
Warranty1 Year (parts and labor)
Unit SizeEach
VO1824CFull specifications
Each127.4L; 208/240V; 1500w; Painted cabinet; 1 door, 2 shelves127.4 LRequest A Quote
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Description127.4L; 208/240V; 1500w; Painted cabinet; 1 door, 2 shelves
Capacity (Metric)127.4 L
Temperature (Metric)6°C to 260°C
TypeDigital, OTP
DisplayLED
MaterialPainted Steel
No. of Shelves2
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
Length (English)24 in.
Length (Metric)61 cm
Width (English)27 in.
Width (English) Interior18 in.
Width (Metric)69 cm
Width (Metric) Interior46 cm
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V
Wattage1500 w
Frequency50/60 Hz
Unit SizeEach
VO1824SAFull specifications
Each127.4L; 120V; 1500w; SS cabinet; 1 door, 2 shelves127.4 LRequest A Quote
Capacity (English)4.5 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior24 in.
Depth (Metric) Interior61 cm
Dimensions (L x W x H)24 x 27 x 35 in. (61 x 69 x 89 cm)
DisplayLED
Frequency50/60 Hz
Height (English)35 in.
Height (English) Interior18 in.
Height (Metric)89 cm
Height (Metric) Interior46 cm
Length (English)24 in.
Length (Metric)61 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)325 lb.
Shipping Weight (Metric)147 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage1500 w
Width (English) Interior18 in.
Width (Metric) Interior46 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Description127.4L; 120V; 1500w; SS cabinet; 1 door, 2 shelves
Capacity (Metric)127.4 L
Dimensions (D x W x H) Interior24 x 18 x 18 in. (61 x 46 x 46 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialStainless Steel
Width (English)27 in.
No. of Shelves2
Width (Metric)69 cm
Unit SizeEach
VO914AFull specifications
Each18.6L; 120V; 750w; Painted cabinet; 1 door, 3 shelves18.6 LRequest A Quote
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description18.6L; 120V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialPainted Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
VO914CFull specifications
Each18.6L; 208/240V; 750w; Painted cabinet; 1 door, 3 shelves18.6 LRequest A Quote
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage208/240 V
Description18.6L; 208/240V; 750w; Painted cabinet; 1 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements208/240 V 50/60 Hz
MaterialPainted Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
VO914SAFull specifications
Each18.6L; 120V; 750w; SS cabinet; 2 door, 3 shelves18.6 LRequest A Quote
Capacity (English)0.65 cu.ft.
Chamber MaterialCorrosion-resistant Stainless Steel
Controller TypeDigital Microprocessor Control
Depth (English) Interior14 in.
Depth (Metric) Interior36 cm
Dimensions (L x W x H)23 x 18 x 26 in. (58 x 46 x 66 cm)
DisplayLED
Frequency50/60 Hz
Height (English)26 in.
Height (English) Interior9 in.
Height (Metric)66 cm
Height (Metric) Interior23 cm
Length (English)23 in.
Length (Metric)58 cm
PortsInert gas port, vacuum port and vent port
Safety FeaturesOvertemperature Safety System
Shipping Weight (English)150 lb.
Shipping Weight (Metric)68 kg
Temperature Resolution±1°C
Temperature Uniformity±3.5°C
Vacuum Range1 x 10-2 torr (10 microns)
Wattage750 w
Width (English) Interior9 in.
Width (Metric) Interior23 cm
Temperature (Metric)6°C to 260°C
Voltage120 V
Certifications/ComplianceUL listed
Description18.6L; 120V; 750w; SS cabinet; 2 door, 3 shelves
Capacity (Metric)18.6 L
Dimensions (D x W x H) Interior14 x 9 x 9 in. (36 x 23 x 23 cm)
TypeDigital, OTP
Electrical Requirements120 V 50/60 Hz
MaterialStainless Steel
Width (English)18 in.
No. of Shelves3
Width (Metric)46 cm
Unit SizeEach
Showing 9 of 9

Single-Setpoint Digital Microprocessor Control

  • Maximum temperature: 260°C (500°F) with nominal uniformity of ±3.5% of setpoint
  • Simultaneous LED display of setpoint and actual temperature with push-button view; resolution to within 1°C
  • Keypad data entry and pull-out control module for easy service
  • Independent overtemperature safety system factory set with reset pushbutton

Vacuum System

  • 1 in.(2.54cm) manifold, chamber rear, exterior, connects to pump or in-house vacuum source
  • With inert gas injection valve, fresh air inlet, vacuum exhaust port, controllable vacuum release vent port
  • 1000 microns (1 torr) capacity
  • Vacuum gauge, vacuum, gas and inlet valves located on front

Construction

  • Corrosion-resistant stainless-steel cabinet includes three aluminum shelves with painted or stainless-steel finish
  • Rear-mounted vacuum manifold connects to pump (not included) or in-house vacuum source
  • Low-watt heating element extends oven life
  • Heavy-gauge steel, dual-wall construction with fiberglass insulation for minimum heat loss in walls and door
  • High-temperature silicone gasket is specially molded for a positive seal
  • Magnetic latch for positive door seal (4.5 cu. ft. capacity models include mechanical latches)
  • Viewing windows: 0.5 in.(1.3cm) thick in two smaller capacity models or 0.75 in.(1.9cm) thick in 4.5 cu. ft. model; permit observation of work in progress without loss of heat
  • Stainless-steel exterior models come with double door—a solid outer door over inner window door

Includes

Power cord and aluminum shelves (number of shelves based on model; see ordering table)

Warranty: 1 year warranty

Figures