Use the Thermo Scientific™ Vanquish™ Duo UHPLC Systems to support four workflows (Dual LC, Tandem LC or LC-MS, Dual LC-MS and Inverse Gradient) by combining two flow paths in one integrated UHPLC solution. These workflows improve your productivity by saving time, reducing cost per sample, increasing capacity without added bench, and using your resources more efficiently. Vanquish Duo UHPLC Systems expand the benefits of the Vanquish platform without compromising performance, robustness, or ease-of-use.
Description | Vanquish Duo UHPLC System for Dual LC-MS Workflows |
For Use With (Application) | Double the throughput of any LC-MS analysis or run two LC-MS applications simultaneously. |
For Use With (Equipment) | Vanquish Horizon or Vanquish Flex Modules |
Type | Integrated, biocompatible UHPLC System |
Max. Pressure | 1500 bar (22,000 psi), depending on configuration |
Unit Size | undefined |
Description | Vanquish Duo UHPLC System for Inverse Gradient LC Workflows |
For Use With (Application) | Improve your lab's quantification capabilities. This system employs the unique and universal charged aerosol detection to quantify any compound even when no reference standard is available. Setup methods with smart Chromeleon CDS wizards. |
For Use With (Equipment) | Vanquish Horizon or Vanquish Flex Modules |
Type | Integrated Biocompatible UHPLC System |
Max. Pressure | 1500 bar (22,000 psi) depending on configuration |
Unit Size | Each |
Description | Vanquish Duo UHPLC System for Dual LC Workflows |
For Use With (Application) | Two channel LC doubles throughput, deepens sample knowledge, saves on bench space, and improves return on investment. Control the system with easy to use Chromeleon CDS. |
For Use With (Equipment) | Vanquish Horizon or Vanquish Flex Modules |
Type | Integrated Biocompatible UHPLC System |
Max. Pressure | 1500 bar (22,000 psi) depending on configuration |
Solvent Channels | 2 channels |
Unit Size | Each |
Description | Vanquish Duo UHPLC System for Tandem LC or LC-MS Workflows |
For Use With (Application) | Increase lab throughput and return on investment for Tandem LC or LC-MS workflows. Provides offline column regeneration and efficiently uses your detector or mass spectrometer and maximizes the output. Setup methods with smart Chromeleon CDS wizards. |
For Use With (Equipment) | Vanquish Horizon or Vanquish Flex Modules |
Type | Integrated Biocompatible UHPLC System |
Max. Pressure | 1500 bar (22,000 psi) depending on configuration |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
VQDUO-DUALLCMS | Vanquish Duo UHPLC System for Dual LC-MS Workflows | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VQDUO-INVERSE | Each | Vanquish Duo UHPLC System for Inverse Gradient LC Workflows | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VQDUO-DUALLC | Each | Vanquish Duo UHPLC System for Dual LC Workflows | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
VQDUO-TANDEM | Each | Vanquish Duo UHPLC System for Tandem LC or LC-MS Workflows | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Double your throughput, deepen your sample knowledge and better utilize your bench space with the Vanquish Duo for Dual LC workflow, a two channel LC, for improved return on investment.
Vanquish Duo for Tandem LC or LC-MSIncrease your lab's throughput and return on investment with the Vanquish Duo for Tandem LC or LC-MS workflow, an instrument for offline column regeneration, to efficiently use your detector or mass spectrometer and maximize the output.
Vanquish Duo for Dual LC-MSDouble the throughput of your LC-MS application with the Vanquish Duo for Dual LC-MS. Run up to two applcations simultaneously with a multi-channel LC while benefiting from the robustness, performance and ease of use build into the Vanquish platform.
Vanquish Duo for Inverse Gradient LCImprove your lab's quantification capabilities with the Vanquish Duo for Inverse Gradient, employing the unique and universal Charged Aerosol Detection, to quantify all non-volatile and semi-volatile compounds even when no reference standard is available.
More productivity with ease