Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Improve your charged aerosol detector's (CAD) quantitative capabilities with Thermo Scientific™ Vanquish™ Inverse Gradient LC systems. While changes in solvent composition across gradient separations can lead to changes in CAD analyte responses, Vanquish Inverse Gradient LC systems eliminate this effect by ensuring the eluent entering the detector always has the same solvent composition across a gradient. The resulting uniform analyte response enables standard-free quantitation.
Type | Integrated HPLC or Biocompatible UHPLC system |
Max. Pressure | 1500 bar (22,000 psi) depending on configuration |
Description | Vanquish Inverse Gradient LC Systems |
For Use With (Application) | Improve your lab's quantification capabilities. This system employs the unique and universal charged aerosol detection to quantify any compound even when no reference standard is available. Setup methods with smart Chromeleon CDS wizards. |
For Use With (Equipment) | Vanquish Horizon, Vanquish Flex, or Vanquish Core Modules |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
VQDUO-INVERSE | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Vanquish Inverse Gradient LC systems
Improve your lab's quantification capabilities with the Vanquish Inverse Gradient systems, employing the unique and universal Charged Aerosol Detector, to quantify all non-volatile and semi-volatile compounds even when no reference standard is available.
More productivity with ease