Orion™ Versa Star pH / ISE Measurement Module
Orion™ Versa Star pH / ISE Measurement Module
Orion™ Versa Star pH / ISE Measurement Module
Orion™ Versa Star pH / ISE Measurement Module
Thermo Scientific™

Orion™ Versa Star pH / ISE Measurement Module

Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module
Have Questions?
Catalog NumberpH Range
VSTAR-ISE-2.000 to 20.000
Catalog number VSTAR-ISE
Price (USD)
1,449.00
Online Exclusive
1,610.00
Save 161.00 (10%)
Each
pH Range:
-2.000 to 20.000
Price (USD)
1,449.00
Online Exclusive
1,610.00
Save 161.00 (10%)
Each
Add pH, ion concentration, mV, ORP and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH / ISE Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star pH/ISE module offers up to six point pH and ISE calibrations and advanced ISE features including incremental techniques (single and double known addition and subtraction, analate addition and subtraction), low range stability and electrode ID with calibration verification.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter's four channels at any time.

Orion Versa Star pH/ISE Measurement Module:

  • Measure pH, ion concentration, mV, relative mV or ORP with temperature
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Perform up to a six point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb or none
  • Incremental techniques include single known addition, double known addition, single known subtraction, double known subtraction, analate addition and analate subtraction
  • Advanced ISE features include non-linear selectable auto-blank, low range stability and electrode ID with calibration verification
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Accuracy (ISE)±0.2 mV or ±0.05 of reading, whichever is greater
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.002 pH
Advanced ISE FunctionsTimed endpoint, linear point to point, non-linear selectable auto-blank, low concentration range stability, incremental techniques including single known addition and subtraction, double known addition and subtraction, single analyte addition and subtraction
Alarm OutputSet point alarm, high/low limit alarm, calibration due alarm
Calibration1 point (mV mode)
Calibration Points1 point relative mV (RmV) mode, 1 point ORP mode
Calibration Points (ISE)1 to 6
Calibration Points (pH)1 to 6
DescriptionOrion VERSA STAR pH/mV/ISE/Temperature Module
Dimensions (L x W x H)12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.)
IP RatingIP-54
IncludesVERSA STAR pH/mV/ISE/Temperature Module
InputsBNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip
Isopotential Point7.000pH (adjustable), 1.000 ISE (adjustable)
Measurement ModespH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
Methods10 per channel with password protection
ModelOrion Versa Star pH/ISE/mV/RmV/ORP/Temperature Measurement Module
Probe TypepH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe (purchase separately)
Range (ISE)0 to 19999 in ppm, M, mg/L, %, ppb or unitless
Range (ORP)±2000.0mV
Range (mV)±2000.0mV
Relative Humidity Range5 to 85 %, non-condensing
Resolution (ISE)Up to 4 significant digits
Resolution (Temperature)0.1°C
Resolution (mV)0.1mV
Resolution (pH)0.1, 0.01, 0.001
StabilitySelectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Operating41°F to 113°F
Temperature (Metric) Operating5°C to 45°C
Temperature (Metric) Range-5°C to 105deg;C
Temperature Probe Calibration1 point temperature offset calibration
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 year module warranty
Weight (English)0.5 lb.
Weight (Metric)0.2 kg
TypepH/ISE Measurement Module
pH Range-2.000 to 20.000
Unit SizeEach