Orion™ Versa Star pH Measurement Module
Orion™ Versa Star pH Measurement Module
Orion™ Versa Star pH Measurement Module
Orion™ Versa Star pH Measurement Module
Thermo Scientific™

Orion™ Versa Star pH Measurement Module

Add pH, mV, ORP and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH Measurement Module
Have Questions?
Catalog number VSTAR-PH
Price (USD)
764.10
Online Exclusive
849.00
Save 84.90 (10%)
Each
Price (USD)
764.10
Online Exclusive
849.00
Save 84.90 (10%)
Each
Add pH, mV, ORP and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star pH Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star pH module offers up to a six point pH calibration with easy to follow onscreen instructions, automatic buffer recognition of NIST, DIN or user-defined buffer groups and calibration editing option to fix individual points without a full recalibration.

Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter's four channels at any time.

Orion Versa Star pH Measurement Module:

  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
  • Module offers a 3-year replacement warranty

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH)±0.002 pH
Alarm OutputSet point alarm, high/low limit alarm, calibration due alarm
CalibrationpH with calibration editing option, relative mV (RmV), ORP and temperature
Calibration Points1 to 6
DescriptionOrion Versa Star pH measurement module
Dimensions (L x W x H)12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.)
IP RatingIP-54
IncludesVersa Star pH measurement module for use with Versa Star Pro meters to measure pH, mV, relative mV, ORP and temperature using ATC probe or manual entry
InputsBNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Isopotential Point7.000pH (adjustable)
Measurement ModespH, mV, relative mV (RmV) or ORP with temperature
Methods10 per channel with password protection
ModelOrion Versa Star pH/mV/RmV/ORP/Temperature Measurement Module
Probe TypepH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (purchase separately)
Range (ORP)±2000.0mV
Range (mV)±2000.0 mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (Temperature)0.1°F/°C
Resolution (mV)0.1mV
Resolution (pH)0.1, 0.01, 0.001
StabilitySelectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Operating41°F to 113°F
Temperature (English) Range22.0°F to 221.0°F
Temperature (Metric) Operating5°C to 45°C
Temperature (Metric) Range-5°C to 105deg;C
Temperature Probe Calibration1 point temperature offset calibration
Temperature SelectionManual or automatic with ATC temperature probe
Warranty3 year module warranty
Weight (English)0.5 lb.
Weight (Metric)0.2 kg
TypepH Measurement Module
pH Range-2.000 to 20.000
Unit SizeEach