Water Level Regulator Kit for Lindberg/Blue M™ Baths
Product Image
Thermo Scientific™
Water Level Regulator Kit for Lindberg/Blue M™ Baths
Use with the Thermo Scientific™ Lindberg/Blue M water baths.
Have Questions?
Catalog NumberType
WB1110WLFC-1Water Level Regulator
Catalog number WB1110WLFC-1
Price (EUR)
-
Request A Quote
Type:
Water Level Regulator
The Thermo Scientific™ Water Regulator Kit is for use with Thermo Scientific™ Lindberg/Blue M Water Baths.
  • Low-water safety cut-off turns bath off if level falls below setpoint
  • Adjustable electric float switch maintains level at preset depth setpoint
Specifications
For Use With (Equipment)WB1110, WB1120, WB1130, WB1140, RWB3220, RWB3220
TypeWater Level Regulator
Unit SizeEach