| WFLOW-64003 | Full specifications | Each | Thermo Scientific™ Vanquish™ Horizon Binary UHPLC System | Thermo Scientific™ Orbitrap™ Exploris™ 240 Mass Spectrometer | Thermo Scientific™ Chromeleon™ CDS 7.3.1, Thermo Scientific™ BioPharma Finder™ 5.0, Thermo Scientific™ Ardia™ Platform | Request A Quote |
Type | Multi-Attribute Method | Mass Spectrometer Interface | Thermo Scientific™ Orbitrap™ Exploris™ 240 Mass Spectrometer | Liquid Chromatography System | Thermo Scientific™ Vanquish™ Horizon Binary UHPLC System | Software | Thermo Scientific™ Chromeleon™ CDS 7.3.1, Thermo Scientific™ BioPharma Finder™ 5.0, Thermo Scientific™ Ardia™ Platform | For Use With (Application) | UHPLC, HRAM MS | Includes | User guide, training, System Performance Evaluation Tests, service and support by a global, trained and dedicated team of Thermo Scientific MAM experts | Unit Size | Each |
|
| WFLOW-64002 | Full specifications | Each | Thermo Scientific™ Vanquish™ Flex Binary UHPLC System | Thermo Scientific™ Orbitrap™ Exploris™ 240 Mass Spectrometer | Thermo Scientific™ Chromeleon™ CDS 7.3.1, Thermo Scientific™ BioPharma Finder™ 5.0, Thermo Scientific™ Ardia™ Platform | Request A Quote |
Type | Multi-Attribute Method | Mass Spectrometer Interface | Thermo Scientific™ Orbitrap™ Exploris™ 240 Mass Spectrometer | Liquid Chromatography System | Thermo Scientific™ Vanquish™ Flex Binary UHPLC System | Software | Thermo Scientific™ Chromeleon™ CDS 7.3.1, Thermo Scientific™ BioPharma Finder™ 5.0, Thermo Scientific™ Ardia™ Platform | For Use With (Application) | UHPLC, HRAM MS | Includes | User guide, training, System Performance Evaluation Tests, service and support by a global, trained and dedicated team of Thermo Scientific MAM experts | Unit Size | Each |
|
| WFLOW-64004 | Full specifications | Each | Thermo Scientific™ Vanquish™ Flex Binary UHPLC System | Thermo Scientific™ Orbitrap™ Exploris™ MX Mass Detector | Thermo Scientific™ Chromeleon™ CDS 7.3.1 | Request A Quote |
Type | Multi-Attribute Method | Mass Spectrometer Interface | Thermo Scientific™ Orbitrap™ Exploris™ MX Mass Detector | Liquid Chromatography System | Thermo Scientific™ Vanquish™ Flex Binary UHPLC System | Software | Thermo Scientific™ Chromeleon™ CDS 7.3.1 | For Use With (Application) | UHPLC, HRAM MS | Includes | User guide, training, System Performance Evaluation Tests, service and support by a global, trained and dedicated team of Thermo Scientific MAM experts | Unit Size | Each |
|
| WFLOW-64005 | Full specifications | Each | Thermo Scientific™ Vanquish™ Horizon Binary UHPLC System | Thermo Scientific™ Orbitrap™ Exploris™ MX Mass Detector | Thermo Scientific™ Chromeleon™ CDS 7.3.1 | Request A Quote |
Type | Multi-Attribute Method | Mass Spectrometer Interface | Thermo Scientific™ Orbitrap™ Exploris™ MX Mass Detector | Liquid Chromatography System | Thermo Scientific™ Vanquish™ Horizon Binary UHPLC System | Software | Thermo Scientific™ Chromeleon™ CDS 7.3.1 | For Use With (Application) | UHPLC, HRAM MS | Includes | User guide, training, System Performance Evaluation Tests, service and support by a global, trained and dedicated team of Thermo Scientific MAM experts | Unit Size | Each |
|