WG1001BX10는 WG1001BOX와 동일하므로 WG1001BOX로 구매해주세요.
NuPAGE™ Bis-Tris Midi Protein Gels, 8%, 1.0 mm
Invitrogen™

NuPAGE™ Bis-Tris Midi Protein Gels, 8%, 1.0 mm

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
WG1001BX1012 + 2-well
WG1002BX1020-well
WG1003BX1026-well
Catalog number WG1001BX10
Price (KRW)
-
Wells:
12 + 2-well
Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Invitrogen NuPAGE Bis-Tris protein gels are precast polyacrylamide gels that provide broad molecular weight protein separation with high resolution and sample integrity. These precast gels are ideal for applications where protein integrity is crucial. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is important.

Features of NuPAGE Bis-Tris gels include:

  • Better protein integrity—optimized sample preparation process preserves proteins
  • Wide ranges of molecular weight separation—select the right gel and running buffer for optimal separation of your proteins
  • Faster run times—get separation of your proteins in as little as 35 minutes
  • Longer shelf life—store for at least 12 months at room temperature

Choose the right NuPAGE Bis-Tris gel for your protein separation

Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. NuPAGE Bis-Tris protein gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and a 4–12% gradient. Gels come in two sizes: mini (8 x 8 cm) or midi (8.7 x 13.3 cm) and either 1.0 mm (mini and midi gels) or 1.5 mm (mini gel format only) in thickness. NuPAGE Bis-Tris gels also come in multiple well formats.

NuPAGE Bis-Tris gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, use the NuPAGE LDS Sample Buffer (Cat. No. NP0007) and NuPAGE Sample Reducing Agent (NP0004). Use NuPAGE Antioxidant (NP0005) in the running buffer to maintain the reduced state of the proteins during the run and to allow maximum band sharpness. The gels can be run using NuPAGE MES SDS Running Buffer (NP0002) to better resolve small proteins or NuPAGE MOPS SDS Running Buffer (NP000102) to resolve medium- to large-size proteins.

NuPAGE Bis-Tris Midi gels can be used with the XCell SureLock Midi Cell gel apparatuses (WR0100) or with the Bio-Rad Criterion Cell using our adapters (WA0999).

For transfer of proteins to a membrane, rapid semi-dry transfer can be done using the Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001). Alternatively, traditional wet transfer can be performed using the Bio-Rad Criterion Cell and NuPAGE Transfer buffer (NP00061).

For Research Use Only.
Specifications
For Use With (Equipment)XCell4 SureLock Midi-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 gels (1 box)
Sample Loading Volume45 μL + 15 μL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Width (Metric)13 cm
Gel Percentage8%
Gel SizeMidi
Gel TypeBis-Tris
Product LineNuPAGE
Separation Range3.5 to 260 kDa
Separation TypeDenaturing
Wells12 + 2-well
Unit Size10 gels (1 box)
Contents & Storage
Store at 4°C to 25°C. Do not freeze.