The Secondary Alkaline-Phoshatase conjugated (Anti-Mouse) Antibody solution is a pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in aRead more
Have Questions?
Catalog Number
Quantity
WP20006
100 mL (2 Bottles)
Catalog number WP20006
Price (USD)
127.28
Special offer
Online exclusive
Ends: 27-Jun-2026
148.00
Save 20.72 (14%)
2 bottles
Quantity:
100 mL (2 Bottles)
Price (USD)
127.28
Special offer
Online exclusive
Ends: 27-Jun-2026
148.00
Save 20.72 (14%)
2 bottles
The Secondary Alkaline-Phoshatase conjugated (Anti-Mouse) Antibody solution is a pre-diluted, ready-to-use formulation containing alkaline phosphatase labeled polyclonal antibodies in a proprietary stabilizing reagent. A goat anti-mouse polyclonal alk-phos conjugate, this Secondary antibody is compatable with both chemiluminescent and chromogenic methods of immunodetection, and will identify antigens bound with a primary mouse antibody.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Primary or SecondarySecondary
ReactivityMouse
Shipping ConditionWet Ice
Whole MoleculeWhole Antibody
ClassificationPolyclonal
ConjugateAlkaline Phosphatase (AP)
FormLiquid
Host SpeciesGoat
IsotypeIgG
Quantity100 mL (2 Bottles)
Target SpeciesMouse
Unit Size2 bottles
Contents & Storage
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
Can I purchase the Secondary Antibody solutions from the WesternBreeze Chemiluminescent Detection kits as standalone products?
The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007), but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.
Can I purchase the Secondary Antibody solutions from the WesternBreeze Chromogenic Detection kits as standalone products?
The goat anti-mouse and goat anti-rabbit secondary antibody solutions AP-conjugated are available as standalone products (Cat. Nos. WP20006 and WP20007) but the rabbit anti-goat secondary antibody solution AP-conjugated is not available as a standalone product.