Niton™ XL3t GOLDD+ XRF Analyzer
Niton™ XL3t GOLDD+ XRF Analyzer
Niton™ XL3t GOLDD+ XRF Analyzer
Niton™ XL3t GOLDD+ XRF Analyzer
Niton™ XL3t GOLDD+ XRF Analyzer
Niton™ XL3t GOLDD+ XRF Analyzer
Thermo Scientific™

Niton™ XL3t GOLDD+ XRF Analyzer

Achieve fast, accurate analysis of light elements without helium purging or vacuum pumps with handheld XRF analyzers.

Have Questions?
Catalog NumberDetector Type
XL3TGOLDDPLUSGeometrically Optimized Large Area Drift Detector
Catalog number XL3TGOLDDPLUS
Price (CLP)
-
Detector Type:
Geometrically Optimized Large Area Drift Detector

Industries from mining and exploration to scrap metal recycling depend on fast, accurate elemental analysis. The Thermo Scientific™ Niton™ XL3t GOLDD+ XRF Analyzer provides reliable, real-time data for sample identification and positive material identification for even difficult-to-analyze light elements, without helium purging or vacuum pumps. The Niton XL3t GOLDD+ Analyzer delivers a feature-rich package enhanced with Geometrically Optimized Large Area Drift Detector (GOLDD™) technology for improved light element (Mg-S) analysis, the lowest limits of detection and fastest measurement times.

Performance:

GOLDD Technology: When low detection limits and high sample throughput are critical, the Niton XL3t GOLDD+ XRF analyzer delivers measurement times up to 10-times faster than conventional Si-PIN detectors and up to 3-times more precise than conventional smaller silicon drift detectors (SDD) - without helium purge or vacuum. For further enhancing light element analysis, a helium purge option is available. Niton analyzers also feature the highest performance X-ray tube and speed detector available on the market.

Wide Range of Applications:

Scrap Metal Recycling:Sorts aluminum, titanium and bronze alloys, and achieves superior performance for tramp and trace element analysis.

Mining Exploration: Fast, accurate analysis of light elements. Identifies anomalies near the averages naturally found in the earth's crust. Facilitates advanced exploration and soil analysis, concentrate measurement, ore blending, precious metal identification and lab analysis prequalification.

Environmental: Detects contaminants such as chlorine and sulfur in sediment and arsenic in soil. Identifies environmental hazards in residential, commercial and industrial settings, such as toxic drywall.

Features

  • Standard analytical range: Up to 30 elements from Mg to U (varies by application).

  • Most powerful x-ray tube on the market for handheld XRF analysis.

  • Point and shoot simplicity- very easy to use even by nontechnical personnel.

  • Rugged design for real-world industrial environments.

  • Nondestructive analysis with near instantaneous results.

  • Sealed against moisture and dust.

  • Ergonomically designed.

  • Daylight-readable icons.

  • Incorporates multi-language options.

  • Color, touch-screen display.

Connectivity:

  • Standard Thermo Scientific™ Niton Data Transfer (NDT™) PC software suite allows you to set operator permissions, generate custom reports, print certificates of analysis, or remotely monitor, and operate the instrument hands-free from your PC.

  • Integrated USB and Bluetooth™ communications provide direct data transfer to your PC or networked storage device. Optional accessories include Bluetooth barcode reader, Bluetooth printer and Bluetooth GPS.

Specifications
Analytical RangeUp to 30 elements from Mg to U (varies by application)
Available Analytical ModesVaries by application:Alloy Modes: Metal Alloy, Electronics Alloy, Precious MetalsBulk Modes: Soil, Mining, TestAllPlastic Modes: RoHS Plastics, Toy and Consumer Goods, Plastics, TestAll, Painted ProductsCustom Modes: Upon request (bas
Certifications/ComplianceCE, RoHS
Data EntryTouch-screen keyboard, user-programmable pick lists, optional wireless remote barcode reader
Data Memory32MB internal system memory/128 MB internal user storage
Data StorageInternal >10,000 readings with spectra
Data TransferUSB, Bluetooth and RS-232 serial communication
DescriptionNiton XL3t GOLDD+ XRF Analyzer
Dimensions (L x W x H)9.6 x 9.05 x 3.75 in. (244 x 230 x 95.5 mm)
Display TypeTilting, color, touch-screen display
Height (English)3.75 in.
Height (Metric)95.5 mm
Length (English)9.6 in.
Length (Metric)244 mm
LicensingVaries by region. Contact your local distributor.
Max. Power2W
Operating SystemOwnership
Optional Accessories3mm small-spot collimation; Portable test stand; stationary (benchtop) test stand; mobile test stand, Field Mate; Thermo Scientific™ Extend-a-Pole™; Welding mask; Thermo Scientific™ HotFoot™ hot surface adapter; Soil testing guard
Power Consumption0 to 200µA
Security FeaturesPassword-protected user security
Standard AccessoriesIntegrated CCD camera for locating and storing images; Locking shielded carrying case; Shielded belt holster; Two lithium-ion battery packs; Spare battery pack; 110/220 VAC battery charger/AC adaptor; PC connection cables (USB and RS-232); Thermo Scientific™ Niton Data Transfer (NDT™) PC software; Safety lanyard; Check samples/standards
System Electronics533 MHz ARM 11 CPU, 300 MHz dedicated DSP, 80 MHz ASICS DSP for signal processing
Tube TypeAg anode 50 kV and 200 μA
Voltage6 to 50 kV
Weight (English)3 lb.
Weight (Metric)1.3 kg
Width (English)9.05 in.
Width (Metric)230 mm
ApplicationsSample Identification, Positive Material Identification
Detector TypeGeometrically Optimized Large Area Drift Detector
Tube Voltage6 to 50 kV
Unit SizeEach