Coomassie Brilliant Blue R-250, Fisher BioReagents
Coomassie Brilliant Blue R-250, Fisher BioReagents
Coomassie Brilliant Blue R-250, Fisher BioReagents
Coomassie Brilliant Blue R-250, Fisher BioReagents
Fisher Bioreagents

Coomassie Brilliant Blue R-250, Fisher BioReagents

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
Packaging:
View all
Glass Bottle
Catalog number YBP10125
HSN 32041299
Price (INR)
7,120.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
7,120.00
Each
Add to cart
Specifications
Chemical Name or MaterialBrilliant Blue R-250
Melting Point174°C
CAS6104-59-2
ChemAlert Storage SymbolGray
Recommended StorageRT
View more
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.