Sensititre™ YeastOne™ YO11 AST Plate
Thermo Scientific™
Sensititre™ YeastOne™ YO11 AST Plate
Simplify yeast susceptibility testing with this plate.
Have Questions?
Catalog NumberQuantity
YO1110/Pk.
Catalog number YO11
Price (MXN)
-
Quantity:
10/Pk.
Simplify yeast susceptibility testing with the Thermo Scientific™ Sensititre™ YeastOne™ YO11 AST Plate including Micafungin and Rezafungin. The Sensititre YeastOne YO11 AST Plate is intended for research use only. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Achieve reliable, consistent results for Candida species with the Sensititre YeastOne YO11 AST Plate.
  • Room-temperature storage eliminates inventory control concerns
  • Individual packaging allows laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • 24-hour incubation time enables timely results for clinicians
For Research Use Only. Not for use in diagnostic procedures.
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeYeastOne Broth (Part No. Y3462)
DescriptionSensititre YeastOne Research Use Only Plate, w/ Micafungin and Rezafungin
Incubation Conditions35°C non-CO2
Incubation Time24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsManual
Transfer Volume20μL
Volume Per Well100μL
Product TypeAST Plate
SpeciesCandida species
Target Organism ClassYeast
Test TypeYeast Susceptibility