Thermo Scientific™
Sensititre™ YeastOne™ YO2IVD AST Plate
Catalog number: YO2IVD
Thermo Scientific™
Sensititre™ YeastOne™ YO2IVD AST Plate
Catalog number: YO2IVD
Simplify yeast testing for Candida sp. and reduce cost per test using the dual isolate Thermo Scientific™ Sensititre™ YeastOne™ YO2IVD AST Plate with Voriconazole. Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Catalog Number
YO2IVD
Unit Size
Each
Description
Sensititre YeastOne IVD Plate, with Voriconazole
Quantity
10/Pk.
Price (USD)
Full specifications
Antimicrobial AgentsVarious
No. per Pack10
0.5 McFarland StandardH2O (Part No. T3339)
Broth TypeYeastOne Broth (Part No. Y3462)
DescriptionSensititre YeastOne IVD Plate, with Voriconazole
Incubation Time24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
Incubation Conditions35°C O2
Isolates Per Plate2
Reading OptionsManual
Product TypeYeastOne AST Plate
SpeciesCandida Species
Volume Per Well100 μL
Transfer Volume20µL
Quantity10/Pk.
Antibiotic CodeVarious
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)
YO2IVDFull specifications
EachSensititre YeastOne IVD Plate, with Voriconazole10/Pk.Request A Quote
Antimicrobial AgentsVarious
No. per Pack10
0.5 McFarland StandardH2O (Part No. T3339)
Broth TypeYeastOne Broth (Part No. Y3462)
DescriptionSensititre YeastOne IVD Plate, with Voriconazole
Incubation Time24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
Incubation Conditions35°C O2
Isolates Per Plate2
Reading OptionsManual
Product TypeYeastOne AST Plate
SpeciesCandida Species
Volume Per Well100 μL
Transfer Volume20µL
Quantity10/Pk.
Antibiotic CodeVarious
Unit SizeEach
Showing 1 of 1
Simplify yeast testing for Candida sp. and reduce cost per test using the dual isolate Sensititre YeastOne YO2IVD AST Plate with Voriconazole.
  • Room-temperature storage eliminates inventory control concerns.
  • Individual packaging allows laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Cost-effective: the dual isolate plate allows two patient samples per plate reducing the cost per test.
  • 24-hour incubation time enables timely results for clinicians. 
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  