Zenon™ Alexa Fluor™ Plus IgG Labeling Reagents
Zenon™ Alexa Fluor™ Plus IgG Labeling Reagents
Zenon™ Alexa Fluor™ Plus IgG Labeling Reagents
Invitrogen™

Zenon™ Alexa Fluor™ Plus IgG Labeling Reagents

Zenon Alexa Fluor Plus Labeling Reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody binding when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or DyeSpeciesColorExcitation/Emission
Z25615Alexa Fluor™ Plus 488HumanGreen494/519 nm
Z25619Alexa Fluor™ Plus 488MouseGreen494/519 nm
Z25616Alexa Fluor™ Plus 594HumanRed590/617 nm
Z25620Alexa Fluor™ Plus 594MouseRed590/617 nm
Z25617Alexa Fluor™ Plus 647HumanFar-red650/671 nm
Z25621Alexa Fluor™ Plus 647MouseFar-red650/671 nm
Catalog number Z25615
Price (MXN)
-
Label or Dye:
Alexa Fluor™ Plus 488
Species:
Human
Color:
Green
Excitation/Emission:
494/519 nm
Streamline therapeutic antibody discovery campaigns with Zenon Alexa Fluor Plus Labeling Reagents to screen for antibody binding. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody binding when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

An effective cell-based screening model is critical for the success of therapeutic antibody discovery campaigns. This serves as an important filter in the early stages of drug development. Identifying antibodies with optimal binding profiles accelerates candidate selection and helps minimize false positives. The new Zenon Alexa Fluor Plus Labeling Reagents are prelabeled Fab fragments directed against the Fc portion of IgG antibodies. They are improved in three ways: superior pan-reactive Fab fragments, optimized bioconjugation, and next-generation fluorophores. These combine to provide superior signal to background, providing more confidence in screening results.

The Zenon Alexa Fluor Plus IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon labeling reagent concentration used. Formation of the Fab–antibody complex occurs in less than 5 minutes. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. The insights provided by these approaches have the potential to accelerate biotherapeutic lead generation and development.

Advantages of Zenon Alexa Fluor Plus IgG Labeling Reagents include:
• Simplicity—No reagent purification is required
• Accuracy—No wash steps or quenchers are needed, maximizing throughput
• Speed—Labeled antibodies are typically ready to use in 5 minutes
• Specificity—Dyes are designed for the specific detection of antibody binding

Preserve primary antibody function and affinity
Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates
If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission519 nm
Excitation494 nm
Excitation/Emission494/519 nm
Flow Cytometer Laser Lines488
For Use With (Application)Antibody binding screening
Labeling TargetIgG
Label or DyeAlexa Fluor™ Plus 488
Product TypeLabeling Reagent
Quantity240 μL
SpeciesHuman
TargetHuman IgG Fc domain
ColorGreen
Label TypeAlexa Fluor
Product LineZenon
Unit Size240 μL
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.