New Zenon pHrodo Deep Red Labeling Reagents are superior in three ways: improved bioconjugation for brighter signal, redesigned protein for robust screening regardless of IgG isotype, and a new, more sensitive fluorophore.
Zenon™ pHrodo™ iFL IgG Labeling Reagents
Zenon™ pHrodo™ iFL IgG Labeling Reagents
Zenon™ pHrodo™ iFL IgG Labeling Reagents
Invitrogen™

Zenon™ pHrodo™ iFL IgG Labeling Reagents

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량종라벨 또는 염료색상
Z25618250 μLHumanpHrodo Deep RedDeep Red
Z25622250 μLMousepHrodo Deep RedDeep Red
Z25612250 μLHumanpHrodo iFL RedRed
Z25611250 μLHumanpHrodo iFL GreenGreen
Z25610250 μLMousepHrodo iFL RedRed
Z25609250 μLMousepHrodo iFL GreenGreen
Z256231.2 mLHumanpHrodo Deep RedDeep Red
Z256141.2 mLHumanpHrodo iFL RedRed
Z256131.2 mLHumanpHrodo iFL GreenGreen
Z256241.2 mLMousepHrodo Deep RedDeep Red
Z256261.2 mLMousepHrodo RedRed
Z256251.2 mLMousepHrodo GreenGreen
카탈로그 번호 Z25618
제품 가격(KRW)
1,440,000
온라인 행사
Ends: 31-Dec-2026
1,694,000
할인액 254,000
250 μL
수량:
250 μL
종:
Human
라벨 또는 염료:
pHrodo Deep Red
색상:
Deep Red
제품 가격(KRW)
1,440,000
온라인 행사
Ends: 31-Dec-2026
1,694,000
할인액 254,000
250 μL
Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
사양
색상Deep Red
설명pHrodo™ Deep Red, 250 μL
검출 방법Fluorescence
여기/방출640/655 nm
용도(애플리케이션)Antibody binding screening
라벨 유형pHrodo™ (pH sensitive)
제품라인pHrodo
제품 유형Labeling Reagent
수량250 μL
종Human
Labeling TargetIgG
라벨 또는 염료pHrodo Deep Red
Unit Size250 μL
구성 및 보관
Store in refrigerator (2°C to 8°C) and protect from light