Thermo Scientific™
Sterilin™ Tubes with Screw Cap
 Related applications:
Clinical & Translational Research
Hold samples ranging from 5 to 10mL in Thermo Scientific™ Sterilin™ Tubes with Screw Cap. Available in chemically resistant polypropylene or virgin polystyrene with a polyethylene screw cap. Supplied nonsterile. Not available in all geographic regions.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Available labeled for sample identification
  • Supplied nonsterile
Manufactured from either:
  • Virgin polystyrene with flat base
  • Chemically resistant polypropylene with a skirted base
Ordering Information:
Product not available in all geographic regions.
Documents
Manuals & protocols