Product Image

Novex™ Power Supply Adapters

The Novex Power Supply Adapters allow you to connect the covered power leads of electrophoresis units to certain power suppliesRead more
Have Questions?
Catalog NumberQuantity
ZA100011 set
Catalog number ZA10001
Price (KRW)
29,000
Online offer
Ends: 31-Dec-2026
33,000
Save 4,000 (12%)
1 set
Quantity:
1 set
Price (KRW)
29,000
Online offer
Ends: 31-Dec-2026
33,000
Save 4,000 (12%)
1 set
The Novex Power Supply Adapters allow you to connect the covered power leads of electrophoresis units to certain power supplies (for example, some power supplies from Bio-Rad Laboratories, Thermo Fisher Scientific).

The following electrophoresis units from Invitrogen have covered power leads to be in compliance with the latest IEC (International Electrotechnical Commission) and EN (European) standards, and reduce the risk of electrical shock due to exposure to bare metal.

• ZOOM IPGRunner Mini-Cell and ZOOM IEF Fractionator
• XCell4 SureLock Midi-Cell
• Mini Gel Tank
• XCell SureLock Mini-Cell (has retractable power leads)

If you are using any of the above electrophoresis units with power supplies that are not designed for use with covered or retractable power leads, you need to use Novex Power Supply Adapters.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 set
Shipping ConditionRoom Temperature
For Use With (Equipment)XCell SureLock Mini, ZOOM IPGRunner Mini, XCell4 SureLock Midi, ZOOM IEF Fractionator, Mini Gel Tank
Product LineNovex
Unit Size1 set
Contents & Storage
Contains 1 set of adapters and 1 Allen wrench

Frequently asked questions (FAQs)

Can I purchase replacement parts for the Mini Blot Module?

Here are the replacement parts we offer for the Mini Blot Module:

- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I purchase replacement parts for the XCell4 SureLock Midi-Cell?

Here are the replacement parts we offer for the XCell4 SureLock Midi-Cell. They can only be ordered by phone:

- XCell4 SureLock Midi-Cell Lid: Cat. No. WR0101
- XCell4 SureLock Midi-Cell Lower Buffer Chamber: Cat. No. WR0102
- XCell4 SureLock Midi-Cell Displacement Dam: Cat. No. WR0106
- XCell SureLock Upgrade kit: Cat. No. WR0112 (contains 1 SureLock Assembly, 1 buffer dam, and 2 cores) Please note that in addition to ordering on the phone by calling Customer Service, this item can also be ordered using the “Quick Order” link (https://www.thermofisher.com/store/quick-order) that can be found at the top right of most pages on our website. The price will display when the item is added to your shopping cart.

Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.

What are the components of the XCell4 SureLock Midi-Cell?

The XCell4 SureLock Midi-Cell includes the lower buffer chamber, lid with cables, two buffer cores, buffer dam, displacement dam, SureLock assembly, and gel knife. For descriptions and images of the various parts, please refer to pages 1-3 of the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/xcell4_midicell_man.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.