Invitrogen™

ZOOM™ IPG Strip pH 5.3-6.3

Catalog number: ZM0015
Invitrogen™

ZOOM™ IPG Strip pH 5.3-6.3

Catalog number: ZM0015
Catalog Number
ZM0015
Unit Size
12 strips
Price (USD)
172.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ZM001512 strips
172.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
ZOOM® IPG Strips allow you to take advantage of oil-free, trouble-free isoelectric focusing (IEF) with the ZOOM® IPGRunner™ System.

You'll get:

• Maximum IEF resolution zooming in on target proteins
• Fast isoelectric focusing of proteins
• Easy handling in the ZOOM® IPGRunner™ System mini-cell format

Each ZOOM® Strip is 7 cm long and contains a thin layer of polyacrylamide gel that includes a linear pH gradient. Strips are available in wide pH ranges (3-10, 4-7, 6-10, and 9-12) for broader analysis and in one-unit narrow pH ranges (4.5-5.5, 5.3-6.3, or 6.1-7.1) for expanding protein separation within a specific region of interest (Figure 1). ZOOM® Strips are individually labeled with unique identifying numbers, pH range, and orientation marks.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
ZOOM IPG Strips
Gel Type
Polyacrylamide
Separation Type
Isoelectric Focusing
Separation of
Protein
Shipping Condition
Dry Ice
Product Line
ZOOM™
Product Type Specs
IPG Strip
pH Range
5.3 to 6.3
Sample Loading Volume
7 cm (Strips)

Contents & Storage

Each pack of ZOOM™ Strips contains 12 strips attached to a tri-fold card. Store at -20°C. Guaranteed stable for one year when properly stored.

Figures

Frequently asked questions (FAQs)

Citations & References