You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Chromatography Syringes
▸
Manual Gas Tight Syringes
▸
Guided Plunger Syringes
SGE™
Guided Plunger Syringes
Extended barrel guides plunger during injection. SGE™ Guided Plunger Syringes are suitable for industrial environments.
Have Questions?
Catalog Number
Volume (Metric)
Connection Type
Product Type
Length (English) Needle
00-2455
also known as 2455
10 μL
Removable
Guided PTFE Tipped Plunger Syringe
2.75 in.
Catalog number
00-2455
also known as 2455
Price (USD)
-
Volume (Metric):
10 μL
Connection Type:
Removable
Product Type:
Guided PTFE Tipped Plunger Syringe
Length (English) Needle:
2.75 in.
Extended barrel guides plunger during injection. SGE™ Guided Plunger Syringes are suitable for industrial environments.
Extended barrel accommodates a reinforced plunger to provide a rugged low volume syringe
Reinforced plunger prevents bending when the plunger is pulled back to the 100% position
Syringe barrels have a scale length of 54.1mm and an O.D. of 8mm
Fixed-needle models can be heated up to 70°C.
Removable-needle models feature a PTFE seal, a spring and nut for leakfree operation and can be heated up to 120°C
All needles are 26s gauge, stainless steel with #2-bevel points, 0.47mm O.D., and are aligned with zero graduation mark on barrel
Specifications
Diameter (English) Outer Needle
0.018 in.
Diameter (Metric) Outer Needle
0.47 mm
Length (English) Needle
2.75 in.
Model
10R-GP-GT
Needle Point Style
Bevel
Product Type
Guided PTFE Tipped Plunger Syringe
Quantity
Ea.
Connection Type
Removable
Needle Gauge
26 G
Volume (Metric)
10 μL
Unit Size
Each