Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™

This product is GMP Manufactured and meets FCC and NF specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
4 L
70 lb.
200 lb.
700 lb.
Packaging:
View all
Poly Pail (Jerrican)
Catalog number 0262-06
Price (USD)
-
Quantity:
70 lb.
Packaging:
Poly Pail (Jerrican)
This product is GMP Manufactured and meets FCC and NF specifications.
This product is GMP Manufactured and meets FCC and NF specifications.
Specifications
CAS7664-38-2
Chemical Name or MaterialOrthophosphoric acid
ColorColorless
Melting Point28°C
Molecular FormulaH3PO4
PackagingPoly Pail (Jerrican)
SynonymPhosphoric acid
GradeNF, FCC
Molecular Weight (g/mol)98
Percent Purity85 to 88%
Quantity70 lb.
Unit SizeEach