Guanidinium hydrochloride, 99.5-101.0%, white powder, GenAR™ for biotechnology, GMP, Macron Fine Chemicals™
Avantor™

Guanidinium hydrochloride, 99.5-101.0%, white powder, GenAR™ for biotechnology, GMP, Macron Fine Chemicals™

This product is GMP Manufactured and meets NF specifications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberdimQuantityPackaging
0507-88
M0507-04
M0507-06
0507-25
0507-F7
0507-F9
Catalog number 0507-88
Price (USD)
-
This product is GMP Manufactured and meets NF specifications
This product is GMP Manufactured and meets NF specifications
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
FormPowder
Melting Point185°C
Molecular Weight (g/mol)95.53 g/mol
Packaging TypePolyethylene pail, screw top
Purity99.5 to 101%
Quantity12 kg
GradeBiotech
Unit SizeEach