Standard RC Dialysis Tubing
Fisher Scientific™

Standard RC Dialysis Tubing

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
086704B
also known as FISHERMIS-086704B-EACH
16 mm
0867024848 mm
Catalog number 086704B
also known as FISHERMIS-086704B-EACH
Price (USD)
-
Diameter (Metric):
16 mm
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications
Capacity (Metric)2 mL
Diameter (English)0.6 in.
For Use With (Application)Some tubing rolls do not contain glycerin and do not require soaking prior to use
Length (English)98.4 ft.
Length (Metric)30 m
MaterialRegenerated Cellulose
Product TypeDialysis Membrane
QuantityEach
Diameter (Metric)16 mm
MWCO12 to 14 kDa
Unit SizeEach