Vacuum Filter Holders, Borosilicate Glass Bases
Fisher Scientific™
Vacuum Filter Holders, Borosilicate Glass Bases
For use with Fisherbrand™ Vacuum Filter Holders
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDiameter (Metric) Inside
0975327A
also known as FISHERMIS-0975327A-6/PK
Sintered Support Base47 mm
09753103Support Base47 mm
0975317Filter Support25 mm
Catalog number 0975327A
also known as FISHERMIS-0975327A-6/PK
Price (USD)
-
Type:
Sintered Support Base
Diameter (Metric) Inside:
47 mm
For use with Fisherbrand™ Vacuum Filter Holders
  • Material: Borosilicate glass
Specifications
Diameter (English) Inside1.85 in.
Diameter (Metric) Inside47 mm
TypeSintered Support Base
For Use With (Equipment)Vacuum Filter Holder
MaterialBorosilicate Glass
Unit SizePack of 6