1,4-Benzenedimethanol, 99%
1,4-Benzenedimethanol, 99%
1,4-Benzenedimethanol, 99%
Thermo Scientific Chemicals

1,4-Benzenedimethanol, 99%

CAS: 589-29-7 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 105280500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-29-7
IUPAC Name[4-(hydroxymethyl)phenyl]methanol
Molecular FormulaC8H10O2
InChI KeyBWVAOONFBYYRHY-UHFFFAOYSA-N
SMILESOCC1=CC=C(CO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Crystalline powder or needles
Melting point116°C to 119°C
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only