Potassium Iodide Powder Pillows
Product Image
Hach Pacific Pty Ltd

Potassium Iodide Powder Pillows

Reagent for several iodometric titration methods.
Have Questions?
Catalog NumberQuantity
107799100/Pk.
Catalog number 107799
Price (USD)
-
Quantity:
100/Pk.
Reagent for several iodometric titration methods. Pack of 100 powder pillows.
    Specifications

  • Method: 10025
  • Method Number 2: 10026
  • Method Number 3: 8161
  • Method Number 4: 8209
  • Number of tests: 100
  • Platform: Powder Pillows
  • Quantity: 100 pk
  • Range: Various
Specifications
Quantity100/Pk.
RangeVarious
TypePowder Pillows
Unit SizeEach