Cyclododecanol, 98+%
Cyclododecanol, 98+%
Cyclododecanol, 98+%
Thermo Scientific Chemicals

Cyclododecanol, 98+%

CAS: 1724-39-6 | C12H24O | 184.32 g/mol
Have Questions?
Catalog NumberQuantityPackaging
110961000100 gGlass bottle
Catalog number 110961000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1724-39-6
IUPAC Namecyclododecanol
Molecular FormulaC12H24O
InChI KeySFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILESOC1CCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Appearance (Form)Crystalline powder or crystals
Melting point76°C to 80°C
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only