Thermo Scientific Chemicals

Cyclohexylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: 111291000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Cyclohexylbenzene, 98%, Thermo Scientific Chemicals

Catalog number: 111291000
100 mL, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
827-52-1
IUPAC Name
cyclohexylbenzene
Molecular Formula
C12H16
InChI Key
IGARGHRYKHJQSM-UHFFFAOYSA-N
SMILES
C1CCC(CC1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
GC
>=97.5 %
Appearance (Form)
Liquid
Refractive index
1.5250 to 1.5270 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text