IsoMet 1000 Precision Cutter
Buehler™
IsoMet 1000 Precision Cutter
Precision sectioning saw designed for cutting various types of materials with minimal deformation
Have Questions?
Catalog NumberQuantity
112180Ea.
Catalog number 112180
Price (USD)
-
Quantity:
Ea.
The IsoMet 1000 is a precision sectioning saw that is designed for cutting various types of materials with minimal deformation. This precision cutting machine is ideal for delicate parts since it only uses gravity fed force. Its great versatility in chucking allows for holding all types of sample shapes and configurations. This IsoMet 1000 provides today’s laboratories with the ability to precision cut virtually any material including brittle or ductile metals, composites, cements, laminates, plastics, electronic devices, and biomaterials.
  • Flexible settings for varied sample preparation
  • RPM ranges from 100-975 to provide cut quality for varied materials
  • Table saw attachment accessory for precision sample alignment
  • Great versatility in holding all types of sample shapes and configurations
  • Sectioning tool capable of cutting virtually any material including brittle or ductile metals, composites, cements, laminates, plastics, electronic devices and biomaterials
  • When equipped with the optional rotating chuck, sectioning times are dramatically reduced
  • Gravity fed cutting force reduces deformation on delicate samples
Specifications
For Use With (Application)Cutting Various Materials with Minimal Deformation
Product LineIsoMet 1000
QuantityEa.
Product TypePrecision Sectioning Saw
Unit SizeEach