Thermo Scientific Chemicals

1,8-Dihydroxyanthraquinone, 95%, Thermo Scientific Chemicals

Catalog number: 114835000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1,8-Dihydroxyanthraquinone, 95%, Thermo Scientific Chemicals

Catalog number: 114835000
500 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
117-10-2
IUPAC Name
1,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular Formula
C14H8O4
InChI Key
QBPFLULOKWLNNW-UHFFFAOYSA-N
SMILES
OC1=CC=CC2=C1C(=O)C1=C(O)C=CC=C1C2=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
Melting point
190°C to 195°C
HPLC
>=94 %
Appearance (Color)
Yellow-green to orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text