2-Fluoroanisole 98%, Thermo Scientific Chemicals
2-Fluoroanisole 98%, Thermo Scientific Chemicals
2-Fluoroanisole 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoroanisole 98%, Thermo Scientific Chemicals

Have Questions?
Catalog number 119301000
Price (USD)
-
Request bulk or custom format
Specifications
Assay Percent Range98%
Beilstein06, II, 169
CAS Max %100.0
CAS Min %97.5
CAS321-28-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only